Skip to Content
Merck
All Photos(1)

Key Documents

Y0000220

Roxithromycin for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Roxithromycin, Erythromycin 9-(-O-[2-methoxyethoxy]methyloxime)

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$189.00

$189.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$189.00

About This Item

Empirical Formula (Hill Notation):
C41H76N2O15
CAS Number:
Molecular Weight:
837.05
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$189.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

roxithromycin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCC1OC(=O)C(C)C(OC2CC(C)(OC)C(O)C(C)O2)C(C)C(OC3OC(C)CC(C3O)N(C)C)C(C)(O)CC(C)C(=N/OCOCCOC)\C(C)C(O)C1(C)O

InChI

1S/C41H76N2O15/c1-15-29-41(10,49)34(45)24(4)31(42-53-21-52-17-16-50-13)22(2)19-39(8,48)36(58-38-32(44)28(43(11)12)18-23(3)54-38)25(5)33(26(6)37(47)56-29)57-30-20-40(9,51-14)35(46)27(7)55-30/h22-30,32-36,38,44-46,48-49H,15-21H2,1-14H3/b42-31+

InChI key

RXZBMPWDPOLZGW-FEMONOMJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Roxithromycin for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Semisynthetic erythromycin derivative. Impairs NADPH oxidase activation and alters translocation of its cytosolic components to the neutrophil membrane.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Bergogne-Bérézin
Presse medicale (Paris, France : 1983), 25(39), 1982-1988 (1996-12-14)
Macrolides, one of the oldest antibiotic classes, are widely used in out-patient, clinics and hospitals. The major improvement in developing newer derivatives concerns pharmacokinetic properties. Increased half-lives, persisting concentrations in tissues, interstitial fluids and macrophages confer upon newer macrolides significant
Ming Xue et al.
Analytical and bioanalytical chemistry, 412(1), 203-222 (2019-12-06)
This study examines an improved and simplified method for solid-phase extraction (SPE), which offers rapid and accurate determination and identification of 44 pharmaceutically active compounds using ultra-performance liquid chromatography (UPLC) and tandem mass spectrometry (MS/MS). The common active compounds include
B Facinelli et al.
Journal of chemotherapy (Florence, Italy), 3 Suppl 1, 33-35 (1991-01-01)
The usefulness of macrolides in treating respiratory infections has been established for over thirty years. Currently, a great deal of interest is being focused on roxithromycin, a new semisynthetic derivative of erythromycin which is more stable than erythromycin under acidic
A Markham et al.
Drugs, 48(2), 297-326 (1994-08-01)
Roxithromycin is a derivative of the macrolide antibacterial erythromycin with in vitro antibacterial activity resembling that of the parent compound. The drug has activity against some Staphylococcus spp., many Streptococcus spp., Moraxella (Branhamella) catarrhalis, Mycoplasma pneumoniae, Legionella pneumophila and Chlamydia
[Bacillary angiomatosis in HIV infection. Update apropos of a case treated with roxithromycin].
M Duong et al.
Annales de medecine interne, 143(2), 107-109 (1992-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service