Skip to Content
Merck
All Photos(1)

Key Documents

W217820

Sigma-Aldrich

Butyl alcohol

≥99.5%, FCC, FG

Synonym(s):

1-Butanol, n-Butanol, Butyl alcohol

Sign Into View Organizational & Contract Pricing

Select a Size

W217820-SAMPLE-K
$56.20
1 KG
$71.40
4 KG
$153.00
20 KG
$485.00

$56.20


Estimated to ship onMay 23, 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
W217820-SAMPLE-K
$56.20
1 KG
$71.40
4 KG
$153.00
20 KG
$485.00

About This Item

Linear Formula:
CH3(CH2)3OH
CAS Number:
Molecular Weight:
74.12
FEMA Number:
2178
Beilstein:
969148
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
02.004
NACRES:
NA.21
grade:
FG
Kosher
biological source:
synthetic
food allergen:
no known allergens

$56.20


Estimated to ship onMay 23, 2025


Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

vapor density

2.55 (vs air)

Assay

≥99.5%

autoignition temp.

649 °F

expl. lim.

11.2 %

refractive index

n20/D 1.399 (lit.)

bp

116-118 °C (lit.)

mp

−90 °C (lit.)

density

0.81 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

balsamic; sweet; whiskey

SMILES string

CCCCO

InChI

1S/C4H10O/c1-2-3-4-5/h5H,2-4H2,1H3

InChI key

LRHPLDYGYMQRHN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F - Pensky-Martens closed cup

Flash Point(C)

35 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service