Skip to Content
Merck
All Photos(1)

Key Documents

U211200

Sigma-Aldrich

DMT-2′Fluoro-dU Phosphoramidite

≥99.0% (reversed phase HPLC)

Synonym(s):

DMT-2′-Fluoro-dU amidite

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$20,920.00
500 G
$93,830.00

$20,920.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$20,920.00
500 G
$93,830.00

About This Item

Empirical Formula (Hill Notation):
C39H46FN4O8P
CAS Number:
Molecular Weight:
748.78
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.51

$20,920.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

biological source

non-animal source (no BSE/TSE risk)

product line

Proligo Reagents

Assay

≥99% (31P-NMR)
≥99.0% (reversed phase HPLC)

form

powder

mol wt

748.78 g/mol

technique(s)

oligo synthesis: suitable

impurities

≤0.3% water content (Karl Fischer)
≤0.5% P(III) Impurities 100-169ppm (31P-NMR)
≤0.5% single Impurity (reversed phase HPLC)
≤3% residual Solvent content

color

white to off-white

λ

conforms (UV/VIS Identity)

suitability

conforms to structure for H-NMR
conforms to structure for LC-MS

nucleoside profile

base: deoxyuridine
base protecting group: none
2' protecting group: fluoro
5' protecting group: DMT
deprotection: fast/standard

storage temp.

−20°C

SMILES string

COc1ccc(cc1)C(OC[C@H]2O[C@H]([C@H](F)[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)N3C=CC(=O)NC3=O)(c4ccccc4)c5ccc(OC)cc5

InChI

1S/C39H46FN4O8P/c1-26(2)44(27(3)4)53(50-24-10-22-41)52-36-33(51-37(35(36)40)43-23-21-34(45)42-38(43)46)25-49-39(28-11-8-7-9-12-28,29-13-17-31(47-5)18-14-29)30-15-19-32(48-6)20-16-30/h7-9,11-21,23,26-27,33,35-37H,10,24-25H2,1-6H3,(H,42,45,46)/t33-,35-,36-,37-,53?/m1/s1

InChI key

HQHQPAYRJJMYQX-DJTPZYMWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

DMT-2′Fluoro-dU Phosphoramidite is a 2′Fluoro phosphoramidite that is used to synthesize oligonucleotides that are more thermally stable and provide increased nuclease resistance.

Its key features are-

  • Can be employed together with DNA or RNA phosphoramidites
  • Recommended deprotection conditions are 8 hours at 55 °C using concentrated ammonia solution, or with AMA for 10 minutes at 65 °C
  • Synthesis of 2′Fluoro oligonucleotides is similar to standard DNA synthesis but requires an elongated coupling time (recommended is 3 minutes compared to 90 seconds for DNA monomers)
  • Consistent lot-to-lot high purity and performance
  • Manufactured under a certified ISO 9001 quality system

Application

DMT-2′Fluoro-dU Phosphoramidite is suitable for use in the synthesis of 2′-fluoro modified oligonucleotide sequences by employing automated solid-phase chemistry.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service