TURB2
Turbidity 2 NTU Calibration Standard - Formazin
certified reference material
Synonym(s):
Turbidity Calibration Standard (2 NTU)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
certified reference material
Quality Level
CofA
certificate is enclosed in each package
manufacturer/tradename
RTC TURB2-100
concentration
in H2O
application(s)
environmental
format
matrix material
storage temp.
2-8°C
Related Categories
General description
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Application
This CRM can be used to calibrate and validate all types of laboratory turbidity meters in the 2 Nephelometric Turbidity Unit (NTU).
Features and Benefits
- Gravimetrically prepared standard, verified and qualified in an ISO/IEC 17025 and ISO 17034 accredited laboratory
- Traceable to the SI and higher order standards from NIST
- Assigned value based on a purity determination by mass balance and gravimetrically prepared value
- Homogeneity assessed in accordance with ISO Guide 35
- Available in 500 mL high-density polyethylene bottle to assure safe delivery
- Accompanied with a certificate of analysis (CoA) having information on the expiry date for an effective audit
- Prepared using deionized water as a solvent
Other Notes
Values of analytes vary lot to lot. Some lots may not contain all analytes.
related product
Product No.
Description
Pricing
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service