Skip to Content
Merck
All Photos(1)

Key Documents

TURB10

Supelco

Turbidity 10 NTU Calibration Standard - Formazin

certified reference material

Synonym(s):

Formazin Turbidity Calibration Standard 10 NTU

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$120.00
500 ML
$174.00
1 L
$281.00

$120.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$120.00
500 ML
$174.00
1 L
$281.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$120.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

certified reference material

CofA

certificate is enclosed in each package

manufacturer/tradename

RTC TURB10-100

concentration

in H2O

application(s)

environmental

format

matrix material

storage temp.

2-8°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

This CRM can be used to calibrate and validate all types of laboratory turbidity meters in the 10 Nephelometric Turbidity Unit (NTU).

Features and Benefits

  • Gravimetrically prepared standard, verified and qualified in an ISO/IEC 17025 and ISO 17034 accredited laboratory
  • Traceable to the SI and higher order standards from NIST
  • Assigned value based on a purity determination by mass balance and gravimetrically prepared value
  • Homogeneity assessed in accordance with ISO Guide 35
  • Available in high-density polyethylene bottles of volumes 100, 500, and 1000 mL to assure safe delivery
  • Accompanied with a certificate of analysis (CoA) having information on the expiry date for an effective audit
  • Prepared using deionized water as a solvent

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

related product

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. i prodotti turb (turb0-turb1-turb10-turb100-turb1000) sono certificati iso 17034?

    1 answer
    1. Yes. These products are certified reference materials following ISO 17034 and ISO/IEC 17025. Unfortunately, TURB0 is not currently offered.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service