Skip to Content
Merck
All Photos(1)

Key Documents

SYNPLE-SC002

Sigma-Aldrich

Synple 2 Automated Synthesizer

Synple

Synonym(s):

Synple 2

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$37,090.00

$37,090.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
1 EACH
$37,090.00

About This Item

MDL number:
UNSPSC Code:
12161503
NACRES:
NA.22

$37,090.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

Quality Level

SMILES string

[s]1cnc(c1c2ccc(cc2)CNC(=O)[C@H]3N(C[C@@H](C3)O)C(=O)[C@@H](NC(=O)CCOCCC(=O)O)C(C)(C)C)C

InChI key

HVPZHRHEHKZOOJ-TYBLODHISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Synple 2 Automated Synthesizer is a benchtop, capsule-based, easy to use automated synthesis device. Simply add the starting material to be transformed as well as a Synple single-use, disposable reagent capsule, and with a single touch of a button, the product is generated, isolated and purified.

Reagent cartridges are available for the following reaction classes:
  • Heterocycle formation
  • Reductive amination
  • PROTAC formation
  • Biotin tags
  • Mitsunobu reaction
  • Boc protection/deprotection
  • Fluorination

Each cartridge contains a scannable chip enabling the standardized reaction sequence to be loaded without any further programming. Key reaction parameters (time, temperature, etc.) can be easily modified.

Learn more about the Synple automated synthesis platform.

Features and Benefits of Synple Platform
  • Time savings - pre-installed protocols have been highly optimized, which obviates the need for the user′s own reaction development and optimization
  • Easy to use - protocols are loaded by simply scanning the cartridge; does not requires any programming knowledge
  • Safety - all chemicals are contained in the system, minimizing the user′s exposure
  • Set-up reaction in less than 5 minutes
  • Small footprint
  • Plug & Play system - simple installation
  • Reaction temperatures from 25 °C - 80 °C
  • Cartridges available for many different reactions
  • Prefilled with all required reagents
  • RFID chip containing optimized reaction protocol

Legal Information

Product of Synple Chem AG

related product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service