Skip to Content
Merck
All Photos(3)

Key Documents

PPB021

Sigma-Aldrich

SSPE Buffer

pH 7.4, pHast Pack, powder

Synonym(s):

1X SSPE Hybridization buffer, 1X SSPE buffer, SSPE Hybridization buffer, Saline-Sodium Phosphate-EDTA Buffer

Sign Into View Organizational & Contract Pricing

Select a Size

20 PKG
$129.00

$129.00


Estimated to ship onMarch 27, 2025


Request a Bulk Order

Select a Size

Change View
20 PKG
$129.00

About This Item

UNSPSC Code:
41105319
NACRES:
NA.25

$129.00


Estimated to ship onMarch 27, 2025


Request a Bulk Order

product line

pHast Pack

Quality Level

form

powder

pH

7.4

Application

Saline-Sodium Phosphate-EDTA is used as a hybridization buffer in Northern and Southern blotting. It is also used as a wash buffer in DNA-microarray technique.

Features and Benefits

  • Ready-to-use buffer
  • No measuring and pH adjusting needed
  • Save time and minimize effort
  • Biological tests: free of DNase, RNase, protease, and nickase
  • Chemical tests: Iron ≤10 ppm, lead ≤5 ppm

Packaging

Foil pouches

Reconstitution

Contents of one pouch, when dissolved in 1000 mL of ultrapure water, will yield a 1× solution containing 150 mM NaCl, 10 mM NaH2PO4, 1 mM EDTA, pH 7.4 at 25 °C. Contents tested to be DNAse, RNAse, Protease, and Nickase free.

Storage and Stability

Store at room temperature. Product may naturally agglomerate but can be simply broken up within the pouch prior to use.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

pHast Pack is a trademark of Sigma-Aldrich Co. LLC

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service