Skip to Content
Merck
All Photos(1)

Key Documents

PHR2608

Supelco

Probenecid

Synonym(s):

Probenecid, p-(Dipropylsulfamoyl)benzoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$223.00

$223.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
500 MG
$223.00

About This Item

Empirical Formula (Hill Notation):
C13H19NO4S
CAS Number:
Molecular Weight:
285.36
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$223.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. P3000000
traceable to USP 1563003

description

Pharmaceutical Secondary Standard; Certified Reference Material

API family

probenecid

form

powder

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

application(s)

pharmaceutical small molecule

storage temp.

2-30°C

SMILES string

CCCN(CCC)S(=O)(=O)c1ccc(cc1)C(O)=O

InChI

1S/C13H19NO4S/c1-3-9-14(10-4-2)19(17,18)12-7-5-11(6-8-12)13(15)16/h5-8H,3-4,9-10H2,1-2H3,(H,15,16)

InChI key

DBABZHXKTCFAPX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How to prepare it for animal IP injection? Is it soluble in water or PBS?

    1 answer
    1. This is an analytical standard and is not intended for use as an injectable in animals. Instructions for solubility preparation are not available.

      Helpful?

  2. What is the solubility of the product Probenecid PHR2608?

    1 answer
    1. The solubility of the product Probenecid includes water (partly miscible), solutions of alkali hydroxides, ethanol (100 mM), acetone, chloroform, sodium hydroxide (1 M), DMSO (100 mM), methanol, dilute acids (practically insoluble), dilute alkalies, and ether (slightly soluble). A stock solution can be stored below -20°C for several months.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service