Skip to Content
Merck
All Photos(1)

Key Documents

PHR2241

Supelco

Melatonin Related Compound A

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

5-Methoxytryptamine, 2-(5-Methoxyindol-3-yl)ethylamine, 3-(2-Aminoethyl)-5-methoxyindole, 5-MOT, 5-Methoxyindole-3-ethanamine, Deacetylmelatonin, NSC 56422

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$811.00

$811.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
50 MG
$811.00

About This Item

Empirical Formula (Hill Notation):
C11H14N2O
CAS Number:
Molecular Weight:
190.24
Beilstein:
145587
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$811.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1380116

API family

melatonin

form

solid

CofA

current certificate can be downloaded

packaging

pkg of 50 mg

mp

121-123 °C (lit.)

application(s)

pharmaceutical small molecule

storage temp.

-10 to -25°C

SMILES string

COc1ccc2[nH]cc(CCN)c2c1

InChI

1S/C11H14N2O/c1-14-9-2-3-11-10(6-9)8(4-5-12)7-13-11/h2-3,6-7,13H,4-5,12H2,1H3

InChI key

JTEJPPKMYBDEMY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Application

Reactant for preparation of:
  • Carboline disaccharide domain of shishijimicin A
  • Melatonin analogs for the reduction of intraocular pressure
  • 5-HT4 receptor ligands
  • inhibitors of sortase A and isocitrate lyase
  • Therapeutic agents for treatment of ischemia/reperfusion (I/R) injury
  • Aurora and epidermal growth factor receptor kinase inhibitors
  • Agents for the treatment of human papillomavirus infection
  • Manzamine analogues for the control of neuroinflammation and cerebral infections
  • Inhibitors of pro-inflammatory cytokines
  • Tacrine-melatonin hybrids as multifunctional agents for alzheimer′s disease

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3413 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service