Skip to Content
Merck
All Photos(1)

Key Documents

PHR1712

Supelco

Phenylephrine Related Compound E

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

1-(3-Hydroxyphenyl)-2-[methyl(phenylmethyl)amino]ethanone hydrochloride, Benzylphenylephrone, 2-[Benzyl(methyl)amino]-1-(3-hydroxyphenyl)ethanone hydrochloride, Benzylphenylephrone hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$893.00

$893.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$893.00

About This Item

Empirical Formula (Hill Notation):
C16H17NO2 · HCl
CAS Number:
Molecular Weight:
291.77
Beilstein:
3739642
UNSPSC Code:
41116107
NACRES:
NA.24

$893.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1533046

API family

phenylephrine

CofA

current certificate can be downloaded

packaging

pkg of 50 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Cl-].N(Cc2ccccc2)(CC(=O)c1cc(ccc1)O)C.[H+]

InChI

1S/C16H17NO2.ClH/c1-17(11-13-6-3-2-4-7-13)12-16(19)14-8-5-9-15(18)10-14;/h2-10,18H,11-12H2,1H3;1H

InChI key

QGHUDAOMXLLADV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3477 in the slot below. This is an example certificate only and may not be the lot that you receive.

related product

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service