![Ergosterol European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/325/720/5a7f4820-2403-4a08-a391-ba8318e34de9/640/5a7f4820-2403-4a08-a391-ba8318e34de9.png)
Ergosterol
![Ergosterol European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/325/720/5a7f4820-2403-4a08-a391-ba8318e34de9/640/5a7f4820-2403-4a08-a391-ba8318e34de9.png)
Synonym(s):
3β-Hydroxy-5,7,22-ergostatriene, 5,7,22-Ergostatrien-3β-ol, Provitamin D2, Ergosterol
Empirical Formula (Hill Notation):
C28H44O
CAS Number:
Molecular Weight:
396.65
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique