Skip to Content
Merck
All Photos(1)

Key Documents

PHR1471

Supelco

Sodium Alginate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Alginic acid sodium salt, Algin, Sodium alginate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$303.00

$303.00


Estimated to ship onMarch 27, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$303.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$303.00


Estimated to ship onMarch 27, 2025


Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1613462

API family

sodium alginate

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

[Na+].[O-]C(=O)C1O[C@H]([C@H]([C@H]([C@@H]1O)O)O)O

InChI

1S/C6H10O7.Na/c7-1-2(8)4(5(10)11)13-6(12)3(1)9;/h1-4,6-9,12H,(H,10,11);/q;+1/p-1/t1-,2-,3-,4?,6+;/m0./s1

InChI key

MSXHSNHNTORCAW-MPGIDXPLSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Sodium Alginate is the sodium salt of alginic acid, extracted from sea weeds. It is generally used as a stabilizer and thickener in the food industry.

Application

Sodium Alginate may be used as a pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations using high-performance liquid chromatography technique.[1]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB5524 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

A validated HPLC assay method for the determination of sodium alginate in pharmaceutical formulations
Awad H, et al.
Journal of Chromatographic Science, 51(3), 208-214 (2012)
Evaluation of sodium alginate for bone marrow cell tissue engineering.
Wang L, et al.
Biomaterials, 24(20), 3475-3481 (2003)

Questions

1–2 of 2 Questions  
  1. What is the M/G ratio of this product? would it be possible to have it in the certificate?

    1 answer
    1. The M/G ratio is not determined for this product. Unfortunately, It is not a test specification that can be provided for the Certificate of Analysis.

      Helpful?

  2. What is the average molecular weight of this product?

    1 answer
    1. The average molecular weight of this product is not determined. In general, molecular weights for sodium alginate can range from 12,000 to 40,000 daltons. This product is not tested for viscosity, thus the average molecular weight is unknown.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service