Skip to Content
Merck
All Photos(1)

Key Documents

PHR1342

Supelco

Sucralose

Sucralose
1 of 1 reviewers received a sample product or took part in a promotion

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Sucralose, 1,6-Dichloro-1,6-dideoxy-β-D-fructofuranosyl-4-chloro-4-deoxy-α-D-galactopyranoside, E955, Trichlorosucrose

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$177.00

$177.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
500 MG
$177.00

About This Item

Empirical Formula (Hill Notation):
C12H19Cl3O8
CAS Number:
Molecular Weight:
397.63
Beilstein:
3654410
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$177.00


In StockDetails


Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0001344
traceable to USP 1623626

API family

sucralose

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@H](O[C@]2(CCl)O[C@H](CCl)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@H]1Cl

InChI

1S/C12H19Cl3O8/c13-1-4-7(17)10(20)12(3-14,22-4)23-11-9(19)8(18)6(15)5(2-16)21-11/h4-11,16-20H,1-3H2/t4-,5-,6+,7-,8+,9-,10+,11-,12+/m1/s1

InChI key

BAQAVOSOZGMPRM-QBMZZYIRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to in-house working standards.
Sucralose is a polar, chlorinated sugar synthesized from saccharose precursor. It is widely used as a sweetener in a number of food and beverage products.[1]

Application

Sucralose may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by various chromatography techniques.[2][3][4]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

A synthetic sweet tastant detectable by humans. Activates T1R2/T1R3 sweet taste receptors on enteroendocrine cells and elicits increased hormonal secretion of glucagon-like peptide-1 and glucose-dependent insulinotrophic peptide.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA1094 in the slot below. This is an example certificate only and may not be the lot that you receive.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluating the taste masking effectiveness of various flavors in a stable formulated pediatric suspension and solution using the [TM="Astree"] electronic tongue
Campbell GA, et al.
Powder Technology, 224(25), 109-123 (2012)
Physicochemical and microbiological stabilities of a sweetened and calorie-free metformin extemporaneous formulation for pediatrics
Alemon-Medina R, et al.
Latin American Journal of Pharmacy, 31(25), 1253-1260 (2012)
Comprehensive analysis of pharmaceutical products using simultaneous mixed-mode (ion-exchange/reversed-phase) and hydrophilic interaction liquid chromatography
Kazarian AA, et al.
Journal of Separation Science, 37(16), 2138-2144 (2014)
Analysis of sucralose and other sweeteners in water and beverage samples by liquid chromatography/time-of-flight mass spectrometry
Ferrer I and Thurman EM
Journal of Chromatography A, 1217(25), 4127-4134 (2010)
Sucralose--an overview of the toxicity data.
H C Grice et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 38 Suppl 2, S1-S6 (2000-07-07)

Questions

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. Indonesia
    • Review 1
    • Votes 0
    5 out of 5 stars.

    Performed good

    This produceted consistent, good and performed oke

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service