Merck
  • Expression and regulation of the tumor suppressor, SEF, during folliculogenesis in humans and mice.
Expression and regulation of the tumor suppressor, SEF, during folliculogenesis in humans and mice.
Reproduction (Cambridge, England) (2014-08-15)
Ela Lutwak, Christopher A Price, Sagit-Sela Abramovich, Shiri Rabinovitz, Irit Granot, Nava Dekel, Dina Ron
ABSTRACT
Similar expression to FGF (Sef or IL17-RD), is a tumor suppressor and an inhibitor of growth factors as well as of pro-inflammatory cytokine signaling. In this study, we examined the regulation of Sef expression by gonadotropins during ovarian folliculogenesis. In sexually immature mice, in situ hybridization (ISH) localized Sef gene expression to early developing oocytes and granulosa cells (GC) but not to theca cells. Sef was also expressed in mouse ovarian endothelial cells, in the fallopian tube epithelium as well as in adipose tissue venules. SEF protein expression, determined by immunohistochemistry (IHC), correlated well with Sef mRNA expression in GC, while differential expression was noticed in oocytes. High Sef mRNA but undetectable SEF protein levels were observed in the oocytes of primary/secondary follicles, while an inverse correlation was found in the oocytes of preantral and small antral follicles. Sef mRNA expression dropped after pregnant mare's serum gonadotropin (PMSG) administration, peaked at 6-8 h after human chorionic gonadotropin (hCG) treatment, and declined by 12 h after this treatment. ISH and IHC localized the changes to oocytes and mural GC following PMSG treatment, whereas Sef expression increased in mural GC and declined in granulosa-lutein cells upon hCG treatment. The ovarian expression of SEF was confirmed using human samples. ISH localized SEF transcripts to human GC of antral follicles but not to corpora lutea. Furthermore, SEF mRNA was detected in human GC recovered from preovulatory follicles. These results are the first to demonstrate SEF expression in a healthy ovary during folliculogenesis. Hormonal regulation of its expression suggests that SEF may be an important factor involved in intra-ovarian control mechanisms.
Product Description
Digoxigenin, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Supelco
Digoxigenin, analytical standard
Pricing and availability is not currently available.
Supelco
p-Toluidine, for spectrophotometric det. of Au, Tl(III), W, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
p-Toluidine, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
p-Toluidine, 99.6%
Pricing and availability is not currently available.
Sigma-Aldrich
Maleic acid, ReagentPlus®, ≥99% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Pricing and availability is not currently available.
Supelco
Maleic acid, qNMR standard, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.
USP
Maleic acid, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.
Supelco
Maleic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.