Skip to Content
Merck
All Photos(1)

Key Documents

PHL80207

g-Strophanthin

phyproof® Reference Substance

Synonym(s):

Acocantherin, Astrobain, Gratibain, Gratus strophanthin, Kombetin, Ouabain

Sign Into View Organizational & Contract Pricing

Select a Size

20 MG
$582.00

$582.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
20 MG
$582.00

About This Item

Empirical Formula (Hill Notation):
C29H44O12
CAS Number:
Molecular Weight:
584.65
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$582.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

SMILES string

O=C1OCC([C@@H]2[C@@]3(C)C[C@@H](O)[C@]4([H])[C@@]5(CO)[C@H](O)C[C@H](O[C@]6([H])O[C@@H](C)[C@H](O)[C@@H](O)[C@H]6O)C[C@@]5(O)CC[C@@]4([H])[C@@]3(O)CC2)=C1

InChI

1S/C29H44O12/c1-13-22(34)23(35)24(36)25(40-13)41-15-8-19(32)28(12-30)21-17(3-5-27(28,37)9-15)29(38)6-4-16(14-7-20(33)39-11-14)26(29,2)10-18(21)31/h7,13,15-19,21-25,30-32,34-38H,3-6,8-12H2,1-2H3/t13-,15-,16+,17+,18+,19+,21+,22-,23+,24+,25-,26+,27-,28+,29-/m0/s1

InChI key

LPMXVESGRSUGHW-HBYQJFLCSA-N

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Oral - Acute Tox. 3 Inhalation - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iñigo Arberas-Jiménez et al.
Scientific reports, 10(1), 17731-17731 (2020-10-22)
Primary amoebic encephalitis (PAM) is a lethal disease caused by the opportunistic pathogen, Naegleria fowleri. This amoebic species is able to live freely in warm aquatic habitats and to infect children and young adults when they perform risk activities in
Lauren E Williamson et al.
Cell, 183(7), 1884-1900 (2020-12-11)
Eastern equine encephalitis virus (EEEV) is one of the most virulent viruses endemic to North America. No licensed vaccines or antiviral therapeutics are available to combat this infection, which has recently shown an increase in human cases. Here, we characterize
Miaoying Shi et al.
Toxicology in vitro : an international journal published in association with BIBRA, 67, 104891-104891 (2020-05-25)
Cardiotoxicity is an important toxicological endpoint for chemical and drug safety assessment. The present study aims to evaluate two stemcell-based in vitro models for cardiotoxicity screening of chemicals. Eleven model compounds were used to evaluate responses of mouse embryonic stem

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service