Skip to Content
Merck
All Photos(1)

Documents

P9672

Supelco

Probucol

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H48O2S2
CAS Number:
Molecular Weight:
516.84
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CC(C)(C)c1cc(SC(C)(C)Sc2cc(c(O)c(c2)C(C)(C)C)C(C)(C)C)cc(c1O)C(C)(C)C

InChI

1S/C31H48O2S2/c1-27(2,3)21-15-19(16-22(25(21)32)28(4,5)6)34-31(13,14)35-20-17-23(29(7,8)9)26(33)24(18-20)30(10,11)12/h15-18,32-33H,1-14H3

InChI key

FYPMFJGVHOHGLL-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Probucol is a diphenolic compound possessing antioxidant and anti-inflammatory properties. It is commercially used as a lipid lowering drug, which helps in reducing the total plasma cholesterol, low density lipoprotein cholesterol (LDL) and high density lipoprotein cholesterol (HDL).

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Enhances selective uptake of HDL-associated cholesteryl esters.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jonathan Watt et al.
Catheterization and cardiovascular interventions : official journal of the Society for Cardiac Angiography & Interventions, 81(4), 698-708 (2012-05-15)
The aim of this study was to assess the efficacy of stent-based delivery of succinobucol alone and in combination with rapamycin in a porcine coronary model. Current drugs and polymers used to coat coronary stents remain suboptimal in terms of
Chang-Jiang Ge et al.
Chinese medical journal, 125(14), 2472-2476 (2012-08-14)
The plasma cystatin C concentration (PcyC) has been demonstrated to have prognostic value in acute coronary syndrome, but the study of PcyC in patients with borderline coronary lesions is limited. Moreover, the effects of atorvastatin and probucol on PcyC and
Eduardo Luiz Gasnhar Moreira et al.
Neuroscience letters, 541, 193-198 (2013-03-09)
Epidemiological studies indicate that high midlife plasma cholesterol levels increases the risk of Alzheimer's disease. Moreover, middle-aged familial hypercholesterolemia (FH) subjects show a particularly high incidence of mild cognitive impairments (MCI). These evidence points to hypercholesterolemia as one of the
Ryusuke Takechi et al.
Clinical and experimental pharmacology & physiology, 40(1), 45-52 (2012-11-22)
Dysfunction of the blood-brain barrier (BBB) is an early pathological feature of vascular dementia and Alzheimer's disease (AD) and is triggered by inflammatory stimuli. Probucol is a lipid-lowering agent with potent anti-oxidant properties once commonly used for the treatment of
Rong Wu et al.
Digestive diseases and sciences, 58(1), 163-171 (2012-08-11)
We sought to evaluate the effects of probucol on steatohepatitis and associated molecular mechanisms in a rat model of nonalcoholic steatohepatitis (NASH) induced by high-fat diet (HFD). Forty male rats weighing 100-120 g were randomly assigned to the following treatments (n = 10

Articles

Lipoproteins package cholesterol for transport in plasma, essential for lipid transport and cellular function in the body.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service