Skip to Content
Merck
All Photos(1)

Key Documents

SML3200

Sigma-Aldrich

MK-0608

≥98% (HPLC)

Synonym(s):

2′-C-Methyl-7-deaza-adenosine, 4-Amino-7-(2-C-methyl-[beta]-D-ribofuranosyl)-7H-pyrrolo[2,3-d]pyrimidine, 7′DMA, 7-(2-C -Methyl-β-D-ribofuranosyl)-7H-pyrrolo[2,3-d ]pyrimidin-4-amine, 7-DMA, 7-Deaza-2’-C-methyladenosine, 7-Deaza-2’-CMA, 7DMA, MK 0608, MK 608, MK-608, MK0608, MK608

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$141.00
25 MG
$449.00

$141.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$141.00
25 MG
$449.00

About This Item

Empirical Formula (Hill Notation):
C12H16N4O4
CAS Number:
Molecular Weight:
280.28
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

$141.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -155 to -185°, c = 1 in methanol

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

[n]2(c3ncnc(c3cc2)N)[C@@H]1O[C@@H]([C@H]([C@]1(O)C)O)CO

InChI

1S/C12H16N4O4/c1-12(19)8(18)7(4-17)20-11(12)16-3-2-6-9(13)14-5-15-10(6)16/h2-3,5,7-8,11,17-19H,4H2,1H3,(H2,13,14,15)/t7-,8-,11-,12-/m1/s1

InChI key

IRZRJANZDIOOIF-GAJNKVMBSA-N

Biochem/physiol Actions

MK-0608 (7DMA; 2′-C-Methyl-7-deaza-adenosine; 7-Deaza-2′-C-methyladenosine) is an orally active antiviral nucleoside analog that inhibits hepatitis C virus (HCV genotype 1b replicon IC50/90 = 0.3/1.3 ?M) and Zika virus (ZIKV strain MR766 IC50 = 1.3-9.6 μM) replication by targeting viral RNA-dependent RNA polymerase. MK-0608 effectively reduces viral load in HCV-infected (0.2-2 mg/kg chimpanzees via daily i.v. or p.o.; 3-50 mg/kg mice b.i.s. p.o.) and ZIKV-infected animals in vivo (50 mg/kg mice via daily p.o.).
Orally active antiviral nucleoside analog that inhibits hepatitis C virus (HCV) and Zika virus (ZIKV) replication in vitro and in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service