Skip to Content
Merck
All Photos(1)

Key Documents

P8119

Supelco

Protein Standard

analytical standard, 80 mg/mL (HSA and gamma-globulins)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701
NACRES:
NA.24
Pricing and availability is not currently available.

biological source

human

Quality Level

grade

analytical standard

form

liquid

contains

0.05% sodium azide as preservative

analyte chemical class(es)

amino acids, peptides, proteins

concentration

80 mg/mL (HSA and gamma-globulins)

technique(s)

gel permeation chromatography (GPC): suitable

application(s)

food and beverages

format

multi-component solution

storage temp.

2-8°C

Application

Protein standard has been used in the determination of protein content of cod trypsin isoenzymes using bio-rad protein assay and ultraviolet-visible light spectrophotometry (UV-Vis).[1]

Other Notes

Human serum albumin and gamma-globulins

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Elucidation of different cold-adapted Atlantic cod (Gadus morhua) trypsin X isoenzymes
Stefansson B, et al.
Biochimica et Biophysica Acta, 1865(1), 11-19 (2017)
Bjarki Stefansson et al.
Biochimica et biophysica acta, 1865(1), 11-19 (2016-10-25)
Trypsins from Atlantic cod (Gadus morhua), consisting of several isoenzymes, are highly active cold-adapted serine proteases. These trypsins are isolated for biomedical use in an eco-friendly manner from underutilized seafood by-products. Our group has explored the biochemical properties of trypsins
Yu-Cheng Chen et al.
Optica, 3(8), 809-815 (2016-08-20)
Indocyanine green (ICG) is the only near-infrared dye approved by the U.S. Food and Drug Administration for clinical use. When injected in blood, ICG binds primarily to plasma proteins and lipoproteins, resulting in enhanced fluorescence. Recently, the optofluidic laser has
Mehdi Alikhani et al.
Journal of proteomics, 162, 141-154 (2017-02-19)
Male infertility accounts for half of the infertility problems experienced by couples. Azoospermia, having no measurable level of sperm in seminal fluid, is one of the known conditions resulting in male infertility. In order to elucidate the complex molecular mechanisms
Jorid T Stuenaes et al.
British journal of pharmacology, 160(1), 116-129 (2010-04-24)
Genetic approaches have documented protein kinase B (PKB) as a pivotal regulator of heart function. Insulin strongly activates PKB, whereas adrenaline is not considered a major physiological regulator of PKB in heart. In skeletal muscles, however, adrenaline potentiates insulin-stimulated PKB

Questions

  1. What is the extinction coefficient for this 80 mg/mL protein standard?

    1 answer
    1. This product is a mixture of human serum albumin (HSA) and human gamma-globulin, the latter of which is essentially all human IgG. The E1% (at 280 nm) values for HSA and human IgG are 14 and 5.3, respectively. The lot-specific extinction coefficient for the mixture can be calculated using the protein concentrations provided the certificate of analysis (CofA)._x000D_
      Sample calculation, for Lot SLCN8399:_x000D_
      https://www.sigmaaldrich.com/certificates/sapfs/PROD/sap/certificate_pdfs/COFA/Q14/P8119-BULKSLCN8399.pdf_x000D_
      The CofA reports an HSA concentration of 4.9 g/dL, and (by difference) a gamma-globulin concentration of 3.3 g/dL. The extinction coefficient at 280 nm for this 8.2 g/dL (82 mg/mL) lot is 86. Here is the calculation: (4.9 X 14) + (3.3 X 5.3) = 86.09, which would round to 86. If the standard were diluted by a factor of 8.2 to give a 1 g/dL or 1% solution, the E1% would be 10.5.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service