Merck
Lasing in blood.
Optica (2016-08-20)
Yu-Cheng Chen, Qiushu Chen, Xudong Fan
ABSTRACT
Indocyanine green (ICG) is the only near-infrared dye approved by the U.S. Food and Drug Administration for clinical use. When injected in blood, ICG binds primarily to plasma proteins and lipoproteins, resulting in enhanced fluorescence. Recently, the optofluidic laser has emerged as a novel tool in bio-analysis. Laser emission has advantages over fluorescence in signal amplification, narrow linewidth, and strong intensity, leading to orders of magnitude increase in detection sensitivity and imaging contrast. Here we successfully demonstrate, to the best of our knowledge, the first ICG lasing in human serum and whole blood with the clinical ICG concentrations and the pump intensity far below the clinically permissible level. Furthermore, we systematically study ICG laser emission within each major serological component (albumins, globulins, and lipoproteins) and reveal the critical elements and conditions responsible for lasing. Our work marks a critical step toward eventual clinical and biomedical applications of optofluidic lasers using FDA approved fluorophores, which may complement or even supersede conventional fluorescence-based sensing and imaging.