Skip to Content
Merck
All Photos(1)

Key Documents

P7184

Supelco

Sigma Pseudo Corpse Scent

drowned victim scent

Sign Into View Organizational & Contract Pricing

Select a Size

50 CAPS
$220.00

$220.00


Estimated to ship onMarch 19, 2025


Request a Bulk Order

Select a Size

Change View
50 CAPS
$220.00

About This Item

MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$220.00


Estimated to ship onMarch 19, 2025


Request a Bulk Order

Quality Level

format

neat

Looking for similar products? Visit Product Comparison Guide

Application

A valuable training aid for water search. It provides a reliable scent source for 30 to 45 minutes, in still or running water, at depths of 1-12 feet.Canine training aids for the detection of corpses.
Sigma pseudo corpse scent may be used as a test material for determining the components present in it using techniques such as a direct injection, headspace solid phase microextraction (SPME), purge and trap, and thermal desorption followed by gas chromatography coupled with mass spectrometry (GC/MS).[1]
Canine training aids for the detection of corpses.

Legal Information

Sigma Pseudo is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comprehensive characterization of commercially available canine training aids
Tipple CA, et al.
Forensic Science International, 242, 242-254 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service