If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf
NIST2387
Peanut butter
NIST® SRM® 2387
Synonym(s):
Ground Peanut Spread
About This Item
grade
certified reference material
Quality Level
packaging
pkg of 3 x 170 g
manufacturer/tradename
NIST®
format
matrix material
Related Categories
General description
SRM 2387_cert
SRM 2387_SDS
Other Notes
Ash, Calories, Carbohydrates, Total Dietary Fiber, Phosphorus (P), Fat (as the sum of fatty acids as triglycerides), Fat (Extractable), Protein, Solids
Amino acids: Alanine, Arginine, Aspartic acid, Cystine, Glutamic acid, Glycine, Histidine, Isoleucine, Leucine, Lysine, Methionine, Phenylalanine, Proline, Serine, Threonine, Tryptophan, Tyrosine, Valine
Biotoxins / Mycotoxins: Aflatoxin B1, Aflatoxin B2, Total Aflatoxins
Lipids and Fatty acids: Arachidic acid, Arachidonic acid, Behenic acid, Eicosadienoic acid (C20:2), Erucic acid, Gondoic acid, Heptadecenoic acid (C17:1), Lignoceric acid, Linoleic acid, Margaric acid, Monounsaturated Fatty acids, Myristic acid, Oleic acid, Palmitic acid, Palmitoleic acid, Polyunsaturated Fatty acids, Saturated Fatty acids, Stearic acid, Vaccenic acid, α-Linolenic acid
Organic Pollutants: Acrylamide
Trace Elements: Calcium (Ca), Copper (Cu), Iron (Fe), Magnesium (Mg), Manganese (Mn), Potassium (K), Sodium (Na), Zinc (Zn)
Vitamins: Niacin (Vitamin B3), Niacinamide (Vitamin B3), Pantothenic acid (Vitamin B5), Pyridoxal (Vitamin B6), Pyridoxamine (Vitamin B6), Pyridoxine (Vitamin B6), Riboflavin (Vitamin B2), Thiamine (Vitamin B1), Total Vitamin B3 as Niacinamide, Total Vitamin B6 as Pyridoxine, a-Tocopherol, γ- + β-Tocopherol, δ-Tocopherol
Matrix Group: Nuts, Soy, Edible Oils and Fats
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
Helpful?
-
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service