Skip to Content
Merck
All Photos(1)

Key Documents

NIST3669

Arsenic species in frozen human urine (elevated levels)

NIST® SRM® 3669

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

packaging

pkg of 5 x 1.5 mL

manufacturer/tradename

NIST®

application(s)

clinical testing

format

matrix material

General description

This Standard Reference Material (SRM) is intended primarily for validating analytical methods and measurements for the determination of arsenic species in human urine. For more information, please refer to the SDS and COA.

SRM 3669_cert

SRM 3669_SDS

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Arsenous acid (AsIII), Arsenic acid (AsV), Monomethylarsonic acid (MMA), Dimethylarsinic acid (DMA), Arsenobetaine (AB), Arsenic (As)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service