Skip to Content
Merck
All Photos(1)

Documents

NIST3250

Serenoa repens (fruit)

NIST® SRM® 3250

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 5 x 6 g

manufacturer/tradename

NIST®

format

matrix material

General description

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Lipids and Fatty acids: Arachidic acid, Arachidic acid, Behenic acid, Behenic acid, Capric acid, Caprylic acid, Caprylic acid, Gondoic acid, Lauric acid, Lauric acid, Lignoceric acid, Lignoceric acid, Linoleic acid, Linoleic acid, Linolenic acid, Margaric acid, Margaric acid, Myristic acid, Myristic acid, Oleic acid, Oleic acid, Palmitic acid, Palmitic acid, Palmitoleic acid, Palmitoleic acid, Pentadecanoic acid (C15:0), Pentadecanoic acid (C15:0), Stearic acid, Stearic acid, Tridecanoic acid (C13:0), Tridecanoic acid (C13:0), Vaccenic acid, Vaccenic acid
Phytochemicals: Campesterol, Phytosterols, Stigmasterol, β-Sitosterol

Matrix Group: Botanicals

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service