Skip to Content
Merck
All Photos(2)

Documents

NIST2152

Urea

NIST® SRM® 2152, combustion calorimetric standard

Synonym(s):

Carbamide, Carbonyldiamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CONH2
CAS Number:
Molecular Weight:
60.06
Beilstein:
635724
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

solid

packaging

pkg of 25 g

manufacturer/tradename

NIST®

mp

132-135 °C (lit.)

density

1.335 g/mL at 25 °C (lit.)

application(s)

clinical testing

SMILES string

NC(N)=O

InChI

1S/CH4N2O/c2-1(3)4/h(H4,2,3,4)

InChI key

XSQUKJJJFZCRTK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Application

Used for the denaturation of proteins and as a mild solubilization agent for insoluble or denatured proteins. Useful for renaturing proteins from samples already denatured with 6 M guanidine chloride such as inclusion bodies. May be used with guanidine hydrochloride and dithiothreitrol (DTT) in the refolding of denatured proteins into their native or active form.

Other Notes

Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service