Skip to Content
Merck
All Photos(1)

Key Documents

NIST1416

Aluminosilicate Glass for Liquidus Temperature

NIST®SRM®

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$1,120.00

$1,120.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
250 G
$1,120.00

About This Item

Linear Formula:
3Al2O3 · 2SiO2
CAS Number:
Molecular Weight:
426.05
UNSPSC Code:
41116107
NACRES:
NA.24

$1,120.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

packaging

pkg of 250 g

manufacturer/tradename

NIST®

application(s)

glass & ceramic

General description

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Alkaline earth aluminosilicate

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service