Skip to Content
Merck
All Photos(1)

Key Documents

M0553

Millipore

Middlebrook ADC Growth Supplement

Enrichment supplement recommended for the cultivation of Mycobacteria

Synonym(s):

Middlebrook Albumin Dextrose Catalase Supplement

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
$113.00

$113.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
1 VIAL
$113.00

About This Item

UNSPSC Code:
41171614
NACRES:
NA.85

$113.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk OrderRequest more information

Quality Level

sterility

sterile (Filtered and Aseptic Handled)

form

liquid

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing
environmental
food and beverages
life science and biopharma
microbiology
veterinary

storage temp.

2-8°C

suitability

mycobacteria

General description

Middlebrook ADC growth supplement is used to isolate a wide variety of Mycobacteria including M. tuberculosis. It contains Bovine albumin, dextrose, and catalase. Bovine albumin neutralizes the toxic products. Dextrose is the fermentable sugar and catalase cleaves the toxic hydrogen peroxides which creates stress to the Mycobacteria cells thus, making it an ideal media for the cultivation of Mycobacterial species

Application

Specialized culture media supplement, designed to isolate and cultivate various Mycobacterial species.

Components

Ingredients: (per vial)
Bovine albumin fraction V 2.5g
Dextrose 1.0g
Catalase 0.0015g
Distilled water 50 ml

Reconstitution

Warm up the refrigerated supplement to 45-50°C. Aseptically add the contents of the vial to 450 ml of sterile Middlebrook 7H9 Broth Base (M0178). Mix well and pour into sterile screw capped tubes.

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nancy Patience Motlalepula Komape et al.
BMC complementary and alternative medicine, 17(1), 128-128 (2017-02-27)
Tuberculosis is an infectious communicable disease and the causative agent of the disease has over the years developed resistance to streamline chemotherapeutic agents with dire consequences and there is a need for development of new and more potent alternatives. Constituents
Joshua N Gustine et al.
Current microbiology, 76(4), 398-409 (2019-01-04)
Cell wall hydrolases are enzymes that cleave bacterial cell walls by hydrolyzing specific bonds within peptidoglycan and other portions of the envelope. Two major sources of hydrolases in nature are from hosts and microbes. This study specifically investigated whether cell

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service