Skip to Content
Merck
All Photos(1)

Key Documents

M0303

Millipore

Middlebrook 7H10 Agar Base

suitable for microbiology, NutriSelect® Plus, for use in isolation and cultivation of Mycobacterium species

Synonym(s):

Mycobacterial culture media

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$375.00

$375.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 04, 2025Details


Request a Bulk Order

Select a Size

Change View
500 G
$375.00

About This Item

UNSPSC Code:
41171609
NACRES:
NA.85

$375.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 04, 2025Details


Request a Bulk Order

Quality Level

sterility

non-sterile

product line

BioChemika

form

powder

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 500 g

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbe id | susceptibility testing: suitable
microbiological culture: suitable

final pH

6.6±0.2 (25 °C)

application(s)

clinical testing
food and beverages
life science and biopharma
veterinary

microbiology

suitability

mycobacteria

General description

Middlebrook 7H10 Agar Base is recommended for isolation, cultivation and sensitivity testing of Mycobacterium tuberculosis. Middlebrook media consists of many inorganic salts, which help, in growth of Mycobacteria. Citric acid formed from sodium citrate helps in retaining inorganic cations in solution. Glycerol supplies carbon and energy. Middlebrook OADC Growth Supplement (M0678) contains oleic acid, bovine albumin, sodium chloride, dextrose and catalase. Oleic acid and other long chain fatty acids are essential for metabolism of Mycobacteria. Some free fatty acids are toxic to Mycobacteria, but albumin binds to those fatty acids and prevents toxic action on Mycobacteria. Dextrose serves as an energy source. Catalase neutralizes toxic peroxides. Malachite green partially inhibits other bacteria.

Application

Middlebrook 7H10 Agar Base is a selective media for isolation, cultivation and sensitivity testing of Mycobacterium tuberculosis. Mycobacteria grow most rapidly in broth media and hence primary isolation of bacterial species can be carried out. It is inexpensive, easy to prepare and can aid in early detection of acid-fast, Mycobacterium species by culture.

Components

Ingredients (g/L)
Ammonium sulfate, 0.50
L-Glutamic acid, 0.50
Monopotassium phosphate, 1.50
Disodium phosphate, 1.50
Sodium citrate, 0.40
Ferric ammonium citrate, 0.04
Magnesium sulfate, 0.025
Calcium chloride, 0.0005
Zinc sulfate, 0.001
Copper sulfate, 0.001
Pyridoxine hydrochloride, 0.001
Biotin, 0.0005
Malachite green, 0.00025
Agar, 15.00

Preparation Note

Suspend 19.47 g of Middlebrook 7H10 Agar Base in 900 ml of distilled water. Add 5 ml of glycerol (Cat. No. 49767). Boil to dissolve the medium completely. Distribute in 180 ml amounts in erlenmeyer flasks and sterilize by autoclaving at 15 lbs. pressure (121°C) for 10 minutes. Cool to 45°C and aseptically add 20 ml of Middlebrook OADC Growth Supplement (Cat. No. M0678). Mix well and pour into screw capped tubes.Note: Keep prepared medium in the dark before and after inoculation.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A biocide-free mineral oil nanoemulsion exhibiting strong bactericidal activity against Mycobacterium immunogenum and Pseudomonas aeruginosa
Chang SC, et al.
International Biodeterioration & Biodegradation, 70, 66-73 (2012)
Agreement of Middle brook 7H10 with Lowenstein Jensen and accuracy of the Sensititre MYCOTB plate using either method as a reference standard for Mycobacterium tuberculosis first line drug susceptibility testing
Ssengooba W, et al.
PLoS ONE, 13(6), e0199638-e0199638 (2018)
Finegold, E.J., et al.
Bailey and Scott's Diagnostic Microbiology (1990)
Hyebin Song et al.
Cell systems, 12(1), 92-101 (2020-11-20)
Machine learning can infer how protein sequence maps to function without requiring a detailed understanding of the underlying physical or biological mechanisms. It is challenging to apply existing supervised learning frameworks to large-scale experimental data generated by deep mutational scanning
Ronald J Rieder et al.
Antimicrobial agents and chemotherapy, 53(11), 4598-4603 (2009-08-26)
Methods currently used for in vitro drug susceptibility testing are based on the assessment of bacterial growth-related processes. This reliance on cellular reproduction leads to prolonged incubation times, particularly for slowly growing organisms such as mycobacteria. A new rapid phenotypic

Articles

Identifying Mycobacterium species crucial due to varying clinical significance; some pathogenic, others not.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service