Skip to Content
Merck
All Photos(1)

Key Documents

L3152

Supelco

LB broth, Miller

Microbiologically tested

Synonym(s):

Luria-Bertani, Lysogeny broth

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
$240.00

$240.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KG
$240.00

About This Item

UNSPSC Code:
12000000
NACRES:
NA.74

$240.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

application(s)

food and beverages

Application

Lysogeny broth is a classic enriched bacterial growth medium that is used for preparation of plasmid DNA and recombinant proteins, especially in E coli.

Components

Contains: tryptone, yeast extract, and sodium chloride

Preparation Note

Solve 25.0 g in 1 litre distilled water. Autoclave 15 minutes at 121°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martha Reyes-Becerril et al.
Fish & shellfish immunology, 78, 392-403 (2018-04-24)
β-Glucans are naturally occurring polysaccharides that are produced by bacteria, fungi and yeast. They are considered immunostimulants in fish acting on non-specific defense mechanism. Yeast-derived glucans from cell wall (Sterigmatomyces halophilus, β-Gluc/Sh) have been used for this purpose in this
Jesús Zepeda-Cervantes et al.
Vaccine, 37(14), 1928-1937 (2019-03-03)
Porcine Circovirus Type 2 (PCV2) is one of the most important pathogens in pigs around the world. PCV2 is a non-enveloped virus and its capsid is formed by a single protein known as open reading frame 2 (ORF2). The aim
Lauren B A Woodruff et al.
Nucleic acids research, 45(3), 1553-1565 (2016-12-23)
Genetic designs can consist of dozens of genes and hundreds of genetic parts. After evaluating a design, it is desirable to implement changes without the cost and burden of starting the construction process from scratch. Here, we report a two-step
Xiang Yin et al.
Experimental and therapeutic medicine, 18(1), 269-277 (2019-07-02)
Platelets primarily participate in hemostasis and antimicrobial host defense. The present study aimed to investigate the effects of leucine-rich repeat flightless-interacting protein-1 (LRRFIP1) on platelet agglutination. The bacterial strain of LRRFIP1 was used to synthesize the recombinant protein and a
Yehou M D Gnopo et al.
Journal of colloid and interface science, 578, 522-532 (2020-06-17)
Recombinantly engineered bacterial outer membrane vesicles (OMVs) are promising vaccine delivery vehicles. The diversity of exogenous antigens delivered by OMVs can be enhanced by induced fusion of OMV populations. To date there are no reports of induced fusion of bacterial

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service