Skip to Content
Merck
All Photos(1)

Documents

L3027

Supelco

LB agar, Miller

Microbiologically tested

Synonym(s):

Luria-Bertani

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
NACRES:
NA.74

Quality Level

application(s)

food and beverages

Application


  • Phenotypic switching in Candida lusitaniae on copper sulfate indicator agar: association with amphotericin B resistance and filamentation: This research explores the use of LB agar, Miller in observing phenotypic changes in Candida lusitaniae, which has implications for understanding fungal resistance mechanisms to antifungal treatments like amphotericin B, significant for researchers in microbiology and pharmaceutical fields focusing on antimicrobial resistance (Miller NS, Dick JD, Merz WG, 2006).

Other Notes

Contains: tryptone, yeast extract, sodium chloride and agar

Preparation Note

Solve 40.0 g in 1 litre distilled water. Autoclave 15 minutes at 121°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Flett et al.
FEMS microbiology letters, 155(2), 223-229 (1997-11-14)
Many streptomycetes, including S. coelicolor A3(2), possess a potent methyl-specific restriction which can present an effective barrier to the introduction of heterologous DNA. We have compared the efficiency of intergeneric conjugal transfer of different types of plasmids to S. coelicolor
Robert E Campbell et al.
Proceedings of the National Academy of Sciences of the United States of America, 99(12), 7877-7882 (2002-06-13)
All coelenterate fluorescent proteins cloned to date display some form of quaternary structure, including the weak tendency of Aequorea green fluorescent protein (GFP) to dimerize, the obligate dimerization of Renilla GFP, and the obligate tetramerization of the red fluorescent protein
F H Yildiz et al.
Proceedings of the National Academy of Sciences of the United States of America, 96(7), 4028-4033 (1999-03-31)
The rugose colony variant of Vibrio cholerae O1, biotype El Tor, is shown to produce an exopolysaccharide, EPSETr, that confers chlorine resistance and biofilm-forming capacity. EPSETr production requires a chromosomal locus, vps, that contains sequences homologous to carbohydrate biosynthesis genes
Ivan Sondi et al.
Journal of colloid and interface science, 275(1), 177-182 (2004-05-26)
The antimicrobial activity of silver nanoparticles against E. coli was investigated as a model for Gram-negative bacteria. Bacteriological tests were performed in Luria-Bertani (LB) medium on solid agar plates and in liquid systems supplemented with different concentrations of nanosized silver

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service