Skip to Content
Merck
All Photos(1)

Key Documents

E5399

Millipore

Endo agar

suitable for microbiology, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$165.00

$165.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
500 G
$165.00

About This Item

UNSPSC Code:
41171606
NACRES:
NA.85

$165.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

pH

7.5±0.2 (25 °C)

application(s)

bioburden testing
cleaning products
cosmetics
environmental
food and beverages
personal care
veterinary
water monitoring

microbiology

suitability

selective and differential for Enterobacter spp.
selective and differential for Escherichia coli
selective and differential for Klebsiella spp.
selective and differential for Proteus spp.
selective and differential for Salmonella spp.
selective and differential for Shigella spp.
selective and differential for coliforms

Components

Components (g/L)
Peptic Digest of Animal Tissue 10.00
Lactose 10.00
Dipotassium Phosphate 3.50
Sodium Sulphite 2.50
Basic Fuchsin 0.50
Agar 15.00

Preparation Note

Suspend 41.5 g of Endo Agar in 1000 ml of distilled water. Boil to dissolve the medium completely. Sterilize by autoclaving at 15 lbs. pressure (121°C) for 15 minutes. Mix well before pouring into sterile petri plates. Note: Basic fuchsin is a potential carcinogen and care should be taken to avoid inhalation of the powdered dye and contamination of the skin.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Greenberg, A..E., et al.
Standard Methods for Examination of Water and Wastewater (1985)
B Björkstén et al.
The Journal of allergy and clinical immunology, 108(4), 516-520 (2001-10-09)
The intestinal microflora is a likely source for the induction of immune deviation in infancy. The purpose of this study was to prospectively relate the intestinal microflora to allergy development in 2 countries differing with respect to the prevalence of

Articles

Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.

An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service