Skip to Content
Merck
All Photos(3)

Documents

D1685

Sigma-Aldrich

Docusate sodium

meets USP testing specifications

Synonym(s):

Docusate sodium salt, AOT, Bis(2-ethylhexyl) sulfosuccinate sodium salt, Sodium bis(2-ethylhexyl) sulfosuccinate, Sulfobutanedioic acid bis(2-ethylhexyl ester) sodium salt, Sulfosuccinic acid bis(2-ethylhexyl) ester sodium salt, ‘Dioctyl’ sulfosuccinate sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H37NaO7S
CAS Number:
Molecular Weight:
444.56
Beilstein:
4117588
EC Number:
MDL number:
UNSPSC Code:
12161902
PubChem Substance ID:
NACRES:
NA.21

Agency

meets USP testing specifications

Quality Level

description

anionic

Assay

99.0-100.5% anhydrous basis

form

solid

mol wt

444.56 g/mol

technique(s)

protein quantification: suitable

application(s)

detection
pharmaceutical (small molecule)

SMILES string

[Na+].CCCCC(CC)COC(=O)CC(C(=O)OCC(CC)CCCC)S([O-])(=O)=O

InChI

1S/C20H38O7S.Na/c1-5-9-11-16(7-3)14-26-19(21)13-18(28(23,24)25)20(22)27-15-17(8-4)12-10-6-2;/h16-18H,5-15H2,1-4H3,(H,23,24,25);/q;+1/p-1

InChI key

APSBXTVYXVQYAB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Docusate sodium (sodium dioctylsulfosuccinate, DOSS) is commonly used as a surfactant, food additive and stool softener.

Application

Docusate sodium (DOSS, AOT) has been used in the preparation of the following:
  • Luminescent ceramics (phosphors).
  • Stock solution during the quantification of DOSS in sea food by using liquid chromatography tandem mass spectrometry (LC-MS/MS).
  • AOT-water-isooctane reverse micelle system
It has also been used in a study to understand its effect on protein bioactivity, when it is co-encapsulates with proteins in polymer based microsphere systems.
Forms reverse micelles in hydrocarbon solvents; Suitable for the solubilization of the major myelin transmembrane proteolipid

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protein bioactivity and polymer orientation is affected by stabilizer incorporation for double-walled microspheres.
Kokai LE, et al.
J. Controlled Release, 141(2), 168-176 (2010)
Determination of dioctylsulfosuccinate in select seafoods using a QuEChERS extraction with liquid chromatography-triple quadrupole mass spectrometry.
Boyd BL, et al.
Laboratory Information Bulletin (2010)
Preparation and characterization of rare-earth-doped Y2O3 luminescent ceramics by the use of reverse micelles.
Graeve OA and Corral JO.
Opt. Mater., 29(1), 24-30 (2006)
Giovanna Longhi et al.
Physical chemistry chemical physics : PCCP, 13(48), 21423-21431 (2011-11-04)
Positively charged supramolecular aggregates formed in vacuo by n AOTNa (sodium bis(2-ethylhexyl)sulfosuccinate) molecules and n(c) additional sodium ions, i.e. [AOT(n)Na(n+n(c))](n(c)), have been investigated by molecular dynamics (MD) simulations for n = 1-20 and n(c) = 0-5. Statistical analysis of physical
Sonali V Kapse et al.
International journal of pharmaceutics, 429(1-2), 104-112 (2012-03-15)
We disclose a self nanoprecipitating preconcentrate (SNP) of tamoxifen citrate (TMX), which forms TMX loaded polymeric nanoparticles, on dilution with aqueous media. SNP comprised TMX, polymer (Kollidon SR) and surfactant/s dissolved in a pharmaceutically acceptable vehicle. Binary surfactant mixtures of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service