Skip to Content
Merck
All Photos(2)

Key Documents

549215

Sigma-Aldrich

2,5-Di(tert-butylperoxy)-2,5-dimethyl-3-hexyne, blend

Synonym(s):

2,5-Dimethyl-2,5-di(tert-butylperoxy)-3-hexyne, blend

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$412.00

$412.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 G
$412.00

About This Item

Linear Formula:
(CH3)3COOC(CH3)2CC(CH3)2OOC(CH3)3
CAS Number:
Molecular Weight:
286.41
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$412.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

composition

active oxygen content, ~5.2 wt. %
calcium carbonate and silica, ~45 wt. %

mp

88 °C (SADT) (lit.)

density

1.26 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)(C)OOC(C)(C)C#CC(C)(C)OOC(C)(C)C

InChI

1S/C16H30O4/c1-13(2,3)17-19-15(7,8)11-12-16(9,10)20-18-14(4,5)6/h1-10H3

InChI key

ODBCKCWTWALFKM-UHFFFAOYSA-N

Application

Free radical polymerization initiator.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Org. Perox. D - Skin Irrit. 2

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service