Skip to Content
Merck
All Photos(1)

Key Documents

D0513

Supelco

Dibucaine

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H29N3O2
CAS Number:
Molecular Weight:
343.46
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
forensics and toxicology
personal care
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

CCCCOc1cc(C(=O)NCCN(CC)CC)c2ccccc2n1

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Potent, long-acting local anesthetic

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jamie Nelsen et al.
Pediatric emergency care, 25(10), 670-673 (2009-10-17)
Dibucaine is considered one of the most potent and consequently toxic amide anesthetics available, and despite withdrawal from the US market as a spinal anesthetic, it remains accessible as an over-the-counter preparation in the United States. Dibucaine exposures in children
A Seelig
Cell biology international reports, 14(4), 369-380 (1990-04-01)
The interaction between lipids and water soluble amphiphiles was investigated by means of a monolayer technique, monitoring the area increase at constant surface pressure. The area increase could be quantitated and binding isotherms at different surface pressures were measured. A
M Laura Parnas et al.
American journal of clinical pathology, 135(2), 271-276 (2011-01-14)
Butyrylcholinesterase (BChE) metabolizes the paralytic succinylcholine. Extended paralysis occurs in people with inherited BChE variants that may be identified by measuring BChE activity with and without the inhibitor dibucaine to calculate a dibucaine number (DN). Accurate phenotyping requires phenotype-specific BChE
Thaïs Souto-Padrón et al.
Parasitology research, 99(4), 317-320 (2006-04-14)
Although local anesthetics (LA) are considered primarily Na+-channel blockers in the past decade, an alternative action of LA as inhibitors of fusion among compartments of the endocytic/exocytic pathways was described. In epimastigote forms of Trypanosoma cruzi, we observed that 50
Jared A Baird et al.
Pharmaceutical research, 29(1), 271-284 (2011-07-26)
Understanding the critical factors governing the crystallization tendency of organic compounds is vital when assessing the feasibility of an amorphous formulation to improve oral bioavailability. The objective of this study was to investigate potential links between viscosity and crystallization tendency

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service