Skip to Content
Merck
All Photos(1)

Key Documents

COND50

Supelco

Conductivity Standard - 50 μmhos/cm

certified reference material

Synonym(s):

50 μmhos/cm Conductivity Calibration Standard

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
$97.60

$97.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 ML
$97.60

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$97.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

100
300

CofA

current certificate can be downloaded

manufacturer/tradename

RTC COND-50-0.500

concentration

in H2O

application(s)

environmental

format

single component solution

storage temp.

2-30°C

General description

This certified reference material (CRM) is produced and certified according to ISO 17034 and ISO/IEC 17025. The specific conductance value of 50.0 ± 0.6 μmhos/ cm is measured using SI and higher order standards from NIST.

All information regarding the use of this CRM including traceability, homogeneity, expiration and safety is available on the certificate of analysis (CoA).

The sample is delivered as a 500 mL whole volume.

Application

Conductivity calibration standard for regular verification of conductivity meters.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service