Skip to Content
Merck
All Photos(1)

Key Documents

BCR693

Human pancreatic Lipase (from pancreatic juice)

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$208.00

$208.00


Estimated to ship onMarch 28, 2025


Request a Bulk Order

Select a Size

Change View
1 EA
$208.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$208.00


Estimated to ship onMarch 28, 2025


Request a Bulk Order

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

application(s)

clinical

format

matrix material

shipped in

dry ice

storage temp.

−20°C

Analysis Note

For more information please see:
BCR693

Legal Information

BCR is a registered trademark of European Commission

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service