Skip to Content
Merck
All Photos(1)

Key Documents

ARK2174

SAFC

Triethanolamine

Pharma Manufacturing

Synonym(s):

2,2′,2′′-Nitrilotriethanol, Tris(2-hydroxyethyl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HOCH2CH2)3N
CAS Number:
Molecular Weight:
149.19
Beilstein:
1699263
EC Number:
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.21

biological source

synthetic

Quality Level

vapor density

5.14 (vs air)

vapor pressure

0.01 mmHg ( 20 °C)

form

liquid

autoignition temp.

600 °F

expl. lim.

8.5 %

refractive index

n20/D 1.485 (lit.)

useful pH range

7.3-8.3

pKa (25 °C)

7.8

bp

190-193 °C/5 mmHg (lit.)

mp

17.9-21 °C (lit.)

solubility

soluble 149 g/L at 20 °C (completely)

density

1.124 g/mL at 25 °C (lit.)

suitability

suitable for manufacturing use

storage temp.

room temp

SMILES string

OCCN(CCO)CCO

InChI

1S/C6H15NO3/c8-4-1-7(2-5-9)3-6-10/h8-10H,1-6H2

InChI key

GSEJCLTVZPLZKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

354.2 °F - closed cup

Flash Point(C)

179 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Imad J Matouk et al.
Biochimica et biophysica acta, 1843(7), 1414-1426 (2014-04-08)
The oncofetal H19 gene transcribes a long non-coding RNA(lncRNA) that is essential for tumor growth. Here we found that numerous established inducers of epithelial to mesenchymal transition(EMT) also induced H19/miR-675 expression. Both TGF-β and hypoxia concomitantly induced H19 and miR-675
Heidi I Chen et al.
Development (Cambridge, England), 141(23), 4500-4512 (2014-11-08)
Identifying coronary artery progenitors and their developmental pathways could inspire novel regenerative treatments for heart disease. Multiple sources of coronary vessels have been proposed, including the sinus venosus (SV), endocardium and proepicardium, but their relative contributions to the coronary circulation
Samuel V Mussi et al.
Pharmaceutical research, 31(8), 1882-1892 (2014-02-14)
To develop a nanostructured lipid carrier (NLC) co-loaded with doxorubicin and docosahexaenoic acid (DHA) and to evaluate its potential to overcome drug resistance and to increase antitumoral effect in MCF-7/Adr cancer cell line. The NLC was prepared by a hot
Francesco Trepiccione et al.
Kidney international, 86(4), 757-767 (2014-05-03)
Almost half of patients receiving lithium salts have nephrogenic diabetes insipidus. Chronic lithium exposure induces AQP2 downregulation and changes in the cellular composition of the collecting duct. In order to understand these pathophysiological events, we determined the earliest lithium targets
Koji Okamoto et al.
The American journal of pathology, 185(7), 1889-1898 (2015-05-20)
Type 2 diabetes mellitus is a leading health issue worldwide. Among cases of diabetes mellitus nephropathy (DN), the major complication of type 2 diabetes mellitus, the nephrotic phenotype is often intractable to clinical intervention and demonstrates the rapid decline of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service