Skip to Content
Merck
All Photos(1)

Key Documents

33729

Sigma-Aldrich

Triethanolamine

puriss. p.a., ≥99% (GC)

Synonym(s):

2,2′,2′′-Nitrilotriethanol, Tris(2-hydroxyethyl)amine

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$179.00
30 KG
Inquire

$179.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
1 L
$179.00
30 KG
Inquire

About This Item

Linear Formula:
(HOCH2CH2)3N
CAS Number:
Molecular Weight:
149.19
Beilstein:
1699263
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

$179.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

vapor density

5.14 (vs air)

Quality Level

vapor pressure

0.01 mmHg ( 20 °C)

grade

puriss. p.a.

Assay

≥99% (GC)

form

viscous liquid

autoignition temp.

600 °F

expl. lim.

8.5 %

impurities

≤0.0001% heavy metals (as Pb)
≤0.1% ethanolamine
≤0.1% water (Karl Fischer)
≤0.5% diethanolamine (GC)

ign. residue

≤0.005% (as SO4)

refractive index

n20/D 1.485 (lit.)

useful pH range

7.3-8.3

pKa (25 °C)

7.8

bp

190-193 °C/5 mmHg (lit.)

mp

17.9-21 °C (lit.)

transition temp

solidification point 20-22 °C

solubility

soluble 149 g/L at 20 °C (completely)

density

1.124 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤1 mg/kg
sulfate (SO42-): ≤20 mg/kg

cation traces

Fe: ≤1 mg/kg

functional group

amine
hydroxyl

SMILES string

OCCN(CCO)CCO

InChI

1S/C6H15NO3/c8-4-1-7(2-5-9)3-6-10/h8-10H,1-6H2

InChI key

GSEJCLTVZPLZKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triethanolamine (TEA) belongs to the family of ethanolamines. It includes the properties of both amines and alcohols.[1]

Application

Triethanolamine may be used as a derivatizing reagent for the determination of boric acid in water samples using gas chromatography-mass spectrometry technique.[2] Addition of triethanolamine to an acidic buffer facilitates the determination of fluoride in inorganic orthophosphates by using a fluoride-selective electrode.[3]

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

354.2 °F - closed cup

Flash Point(C)

179 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fast quantitative analysis of boric acid by gas chromatography-mass spectrometry coupled with a simple and selective derivatization reaction using triethanolamine
Zeng M-L, et al.
Journal of the American Society For Mass Spectrometry, 21(3), 482-485 (2010)
Triethanolamine as an Efficient and Reusable Base, Ligand and Reaction Medium for Phosphane-Free Palladium-Catalyzed Heck Reactions.
Li HJ and Wang L.
European Journal of Organic Chemistry, 22, 5099-5102 (2006)
Toxicology of mono-, di-, and triethanolamine
Knaak BJ, et al.
Reviews of Environmental Contamination and Toxicology, 74(1-86) (1997)
Final report on the safety assessment of triethanolamine, diethanolamine, and monoethanolamine.
Journal of the American College of Toxicology, 2(7), 183-235 (1983)
Yuri L Lyubchenko et al.
Methods (San Diego, Calif.), 47(3), 206-213 (2008-10-07)
This paper describes protocols for studies of structure and dynamics of DNA and protein-DNA complexes with atomic force microscopy (AFM) utilizing the surface chemistry approach. The necessary specifics for the preparation of functionalized surfaces and AFM probes with the use

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service