Skip to Content
Merck
All Photos(4)

Key Documents

P2308

Sigma-Aldrich

Proteinase K from Tritirachium album

Proteinase K from Tritirachium album

lyophilized powder, BioUltra, ≥30 units/mg protein, for molecular biology

Synonym(s):

Endopeptidase K

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$80.30
10 MG
$98.20
25 MG
$122.00
100 MG
$242.00
500 MG
$1,070.00
1 G
$2,060.00

$80.30


Estimated to ship onApril 11, 2025

Diagnostics Manufacturers should use alternative grade product: SRE0005

Request a Bulk Order

Select a Size

Change View
5 MG
$80.30
10 MG
$98.20
25 MG
$122.00
100 MG
$242.00
500 MG
$1,070.00
1 G
$2,060.00

About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
32160410
NACRES:
NA.54

$80.30


Estimated to ship onApril 11, 2025

Diagnostics Manufacturers should use alternative grade product: SRE0005

Request a Bulk Order

grade

for molecular biology

Quality Level

product line

BioUltra

form

lyophilized powder

specific activity

≥30 units/mg protein

mol wt

28.93 kDa

composition

Protein, ≥90% biuret

impurities

≤1 ppm DNA

foreign activity

DNAse, Nickase and RNAse, none detected

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Useful for the proteolytic inactivation of nucleases during the isolation of DNA and RNA.
Useful for the proteolytic inactivation of nucleases during the isolation of DNA and RNA.
Removes endotoxins that bind to cationic proteins such as lysozyme and ribonuclease A.
Reported useful for the isolation of hepatic, yeast, and mung bean mitochondria
Determination of enzyme localization on membranes
Treatment of paraffin embedded tissue sections to expose antigen binding sites for antibody labeling.
Digestion of proteins from brain tissue samples for prions in Transmissible Spongiform Encephalopathies (TSE) research.

Biochem/physiol Actions

Proteinase K is a stable and highly reactive serine protease. Evidence from crystal and molecular structure studies indicates the enzyme belongs to the subtilisin family with an active-site catalytic triad (Asp39-His69-Ser224). It is stable in a broad range of environments: pH, buffer salts, detergents (SDS), and temperature. In the presence of 0.1-0.5% SDS, proteinase K retains activity and will digest a variety of proteins and nucleases in DNA preparations without compromising the integrity of the isolated DNA.

Unit Definition

One unit will hydrolyze urea-denatured hemoglobin to produce color equivalent to 1.0 μmole of tyrosine per min at pH 7.5 at 37 °C (color by Folin-Ciocalteu reagent).

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mamoru Oshiki et al.
Frontiers in microbiology, 9, 830-830 (2018-05-15)
Detection and genotyping of pathogenic RNA viruses in human and environmental samples are useful for monitoring the circulation and prevalence of these pathogens, whereas a conventional PCR assay followed by Sanger sequencing is time-consuming and laborious. The present study aimed
Mark F Sabbagh et al.
eLife, 7 (2018-09-07)
Vascular endothelial cell (EC) function depends on appropriate organ-specific molecular and cellular specializations. To explore genomic mechanisms that control this specialization, we have analyzed and compared the transcriptome, accessible chromatin, and DNA methylome landscapes from mouse brain, liver, lung, and
Margarita Zayas et al.
PLoS pathogens, 12(1), e1005376-e1005376 (2016-01-05)
Hepatitis C virus (HCV) nonstructural protein (NS)5A is a RNA-binding protein composed of a N-terminal membrane anchor, a structured domain I (DI) and two intrinsically disordered domains (DII and DIII) interacting with viral and cellular proteins. While DI and DII
Sreenivas Adurthi et al.
Scientific reports, 7(1), 17289-17289 (2017-12-13)
Oestrogen controls Foxp3 expression in regulatory T cells (Treg cells) via a mechanism thought to involve oestrogen receptor alpha (ERα), but the molecular basis and functional impact of ERα signalling in Treg cells remain unclear. We report that ERα ligand
Kumiko Muta et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 31(10), 1615-1623 (2016-05-18)
Although histone acetylation, an epigenetic modification, has been reported to be related to the progression of various diseases, its involvement in nephrosclerosis is unclear. Dahl salt-sensitive rats were used as a model of nephrosclerosis in this study. The rats were

Articles

Proteinase K aids in molecular biology applications by digesting structural proteins, removing nucleases, and isolating intact genomic DNA.

Pro K aids in disrupting cell membranes for DNA release, crucial for downstream molecular biology techniques.

Balancing Proteinase K cost with quality and technical support ensures optimal enzyme selection for diverse applications.

Guidelines on use of proteinase K, an enzyme commonly used to degrade proteins, and protect DNA and RNA from degradation in samples.

See All

Protocols

The GenElute™ Blood Genomic DNA Kit Protocol provides a simple and convenient way to isolate pure genomic DNA from fresh or aged whole blood.

The GenElute Mammalian Genomic DNA Purification Kit Protocol describes the isolation of pure, high molecular weight DNA from a variety of mammalian sources.

Proteinase K activity measured via spectrophotometry using hemoglobin substrate, crucial for enzyme characterization.

GenElute™ Bacterial Genomic DNA Kit protocol describes a simple and convenient way for the isolation of pure genomic DNA from bacteria.

Questions

1–10 of 10 Questions  
  1. Hi, I'd like to know what happens if the lyophilized proteinase K (P238) is not stored at -20C for at least a week.

    1 answer
    1. The decline in activity over time due to storage conditions has not been determined. Other Proteinase K products may ship at ambient temperature, indicating little or no degradation of the material is expected. However, this items ships on ice. Maintaining the shipping and storage temperature at -20°C is recommended for this product in order to assure optimum performance.

      Helpful?

  2. Is P2308 designed for antigen unmasking during IHC, and is there literature supporting this application?

    1 answer
    1. Proteinase K has not been tested for use with IHC. Product P2308 is specifically tested for use in molecular biology, particularly for preparing cell lysates and releasing nucleic acids, as well as for PCR applications. While the enzyme has been used to digest hair (keratin) and may have other applications, there are no guarantees that P2308 would be suitable for antigen retrieval. No literature references stating that Proteinase K is commonly used for IHC were found.

      Helpful?

  3. Can P2308 be dissolved in water for nucleic acid preparation? If so, at what concentration? Thanks!

    1 answer
    1. This product may be dissolved in water for nucleic acid preparations at 1 mg/mL.

      Please see the link below to review the product datasheet which includes preparation instructions:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/116/320/p2308dat.pdf

      Helpful?

  4. How to prepare a solution of 20 mg/ml of pK (P2308-5MG)? In the section "Preparation Instructions" it mentions that I need to solve 1 mg in 1 ml of water. Does it means I'll have a final concentration of only 1 mg/ml?

    1 answer
    1. The stock may also be prepared at 20 mg/ml concentration in 50 mM Tris-HCl, pH 8.0, containing 1 mM calcium chloride. This solution is stable for at least 12 months at 4°C. For additional information, please refer to the below publications where the product P2308 has been used.
      1) https://www.nature.com/articles/s41598-017-17102-w#Sec9
      2) https://www.frontiersin.org/articles/10.3389/fmicb.2018.00830/full#h3
      3) https://elifesciences.org/articles/36187#s4

      Helpful?

  5. How to prepare Proteinase K solution to use in antigen retrieval of formalin fixed paraffin embedded tissue sections for the purpose of paraffin immunoflorescence? Which is the diluent of choice to prepare the solution from powder form of proteinase K.

    1 answer
    1. This product is not tested for use in antigen retrieval. The product 21627, IHC Select Proteinase K protocol may be applied to this product, P2308. To prepare a stock solution, dissolve Proteinase K, P2308, at 0.2mg/mL in 0.05M Tris solution, pH 7.5.
      For antigen retrieval in FFPA tissue sections (deparaffinized) the recommended dilution of the stock solution is 1:30.
      End users would need to optimize final dilution

      Helpful?

  6. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  7. What is the recommended pH range for Product P2308, Proteinase K?

    1 answer
    1. Proteinase K is normally used in the pH range of 7.5-8.0.

      Helpful?

  8. How should I store solutions of Product P2308, Proteinase K?

    1 answer
    1. Stock solutions in 40% glycerol of at least 20 mg/ml can be stored in 10 mM Tris-HCl buffer, pH 7.5.with 1 mM  calcium acetate (see product P4850). Solutions are also stable at pH 8 at 4°C.

      Helpful?

  9. What amount of Product P2308, Proteinase K, should I use for nucleic acid preparations?

    1 answer
    1. Amounts for typical usage is 50-200 μg/mL.

      Helpful?

  10. How should I prepare solutions of Product P2308, Proteinase K?

    1 answer
    1. Solutions can be prepared in 25-50 mM Tris-HCl buffer, pH 8.0, containing 1-5 mM Calcium chloride.

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service