Key Documents
Z677280
Dermatril nitrile gloves
size L
Synonym(s):
Dermatril Gloves, Nitrile Gloves, Nitrile Safety Gloves, disposable glove
About This Item
UNSPSC Code:
46181504
NACRES:
NB.35
Pricing and availability is not currently available.
material
blue gloves
blue nitrile
size
L
sterility
non-sterile
feature
CE compliant
latex free
powder-free
packaging
pack of 100 ea (boxes of 100)
manufacturer/tradename
KCL 740
L × thickness
250 mm × 0.11 mm
General description
Dermatril is a blue nitrile, powder-free, disposable glove providing a high level of protection against chemical and bacteriological contamination. The Dermatril disposable glove also displays very good physical durability in comparison to conventional disposable gloves.
All Dermatril disposable gloves are INDIVIDUALLY marked with the appropriate standards mark applied on the inside using a food-friendly dye.
Features and Benefits
  • Anti-static.
  • Free from allergenic substances such as thiurames, benzothiazoles and thiourea.
  • Roughened fingertips for better grip.
  • Rolled cuff edge for easier donning.
  • CE compliant.
  • Meet standards: EN 374 (cat. III), EN 388 (cat. II), EN 455, EN 61340-5-1, ASTM 1671:2001
Legal Information
Dermatril is a trademark of KCL GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)