Skip to Content
Merck
All Photos(1)

Key Documents

95197

Supelco

Kojic acid

analytical standard

Synonym(s):

2-Hydroxymethyl-5-hydroxy-γ-pyrone, 5-Hydroxy-2-hydroxymethyl-4H-4-pyranone

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$144.00

$144.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$144.00

About This Item

Empirical Formula (Hill Notation):
C6H6O4
CAS Number:
Molecular Weight:
142.11
Beilstein:
120895
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$144.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

152-155 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

OCC1=CC(=O)C(O)=CO1

InChI

1S/C6H6O4/c7-2-4-1-5(8)6(9)3-10-4/h1,3,7,9H,2H2

InChI key

BEJNERDRQOWKJM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Kojic acid, a tyrosinase inhibitor, is produced by several fungi species, including Aspergillus oryzae.[1] It exhibits skin-lightening property, due to its ability to chelate the copper-containing enzyme tyrosinase in the formation of melanin.[2] Kojic acid is also commonly used as a whitening agent in regular cosmetics, bleaching cosmetics, etc.[3] It is found to be a therapeutic agent and helps in preventing pigmentation.[3]

Application

Kojic acid may be used as an analytical reference standard for the determination of the analyte in skin whitening cosmetics[2][1] and bleaching cosmetics[3] using high performance liquid chromatography with UV detection (HPLC-UV).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Tyrosinase inhibitor.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of magnesium ascorbyl phosphate, ascorbyl glucoside, kojic acid, arbutin and hydroquinone in skin whitening cosmetics by HPLC
Huang C-S, et al.
Journal of food and drug analysis, 12(1) (2004)
Combining high-performance liquid chromatography with on-line microdialysis sampling for the simultaneous determination of ascorbyl glucoside, kojic acid, and niacinamide in bleaching cosmetics.
Lin CH et al
Analytica Chimica Acta, 581(1), 102-107 (2007)
Quantitative determination of $\alpha$-arbutin, $\beta$-arbutin, kojic acid, nicotinamide, hydroquinone, resorcinol, 4-methoxyphenol, 4-ethoxyphenol, and ascorbic acid from skin whitening products by HPLC-UV
Wang YH, et al.
Journal of AOAC (Association of Official Analytical Chemists) International, 98(1), 5-12 (2015)
Atsushi Yoshimori et al.
Bioorganic & medicinal chemistry, 22(21), 6193-6200 (2014-10-08)
Tyrosinase inhibitors have become increasingly critical agents in cosmetic, agricultural, and medicinal products. Although a large number of tyrosinase inhibitors have been reported, almost all the inhibitors were unfortunately evaluated by using commercial available mushroom tyrosinase. Here, we examined the
Safak Ozhan Kocakaya et al.
Iranian journal of pharmaceutical research : IJPR, 19(2), 187-198 (2020-11-24)
Recently Nutrition and Food Chemistry researches have been focused on plants and their products or their secondary metabolites having anti-alzheimer, anti-cancer, anti-aging, and antioxidant properties. Among these plants Salvia L. (Lamiaceae) species come into prominence with their booster effects due

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service