Skip to Content
Merck
All Photos(1)

Documents

938807

Sigma-Aldrich

Potassium fluoride ChemBeads

new

Synonym(s):

KF, Potassium monofluoride, Potassium monofluoride (KF)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
KF
CAS Number:
Molecular Weight:
58.10
MDL number:
UNSPSC Code:
12352100

form

solid

Quality Level

composition

loading of base, 14-16 wt. %

SMILES string

[F-].[K+]

InChI

1S/FH.K/h1H;/q;+1/p-1

InChI key

NROKBHXJSPEDAR-UHFFFAOYSA-M

General description

Potassium fluoride is a white crystalline solid that is highly soluble in water. It is a strong base widely used in the field of catalysis, polymer synthesis, and semiconductor devices. KF has a relatively high melting point. It dissolves well in hydrofluoric acid. However, KF is insoluble in alcohol.

Application

Potassium fluoride can be used:
- To fabricate CIGS thin films solar cells. Efficiency increased by up to 21.7% after KF postdeposition treatment. This was due to the formation of a K-containing layer on the CIGS surface, which resulted in increased surface reactivity due to the formation of a bilayer structure in which a K—In—Se species covers the CIGSe compound.
- To fabricate 2D vanadium carbide MXene for supercapacitor electrodes.
- To synthesize sulfonyl fluoride through electrochemical oxidative coupling.
- As an electrolyte additive for Li-ion batteries.
- As a catalyst for ring-opening fluorination of cyclic thioethers.
- To synthesize solid base catalysts, KF/Ca−Mg−Al hydrotalcite for the production of Biodiesel via transesterification reactions.
A versatile fluoride ion source used in a study of ion-specific swelling and de-swelling of ampholytic polymer gels, in the room temperature oxidation of noble metals in HF, and in the measurement of electronic polarizabilities of ions in polymers of alkali halides.
ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.
For general uses, product is also available in powdered form (449148)

related product

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service