Skip to Content
Merck
All Photos(1)

Key Documents

93453

Supelco

Triclosan

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

5-Chloro-2-(2,4-dichlorophenoxy)phenol, Irgasan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H7Cl3O2
CAS Number:
Molecular Weight:
289.54
Beilstein:
2057142
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

SMILES string

Oc1cc(Cl)ccc1Oc2ccc(Cl)cc2Cl

InChI

1S/C12H7Cl3O2/c13-7-1-3-11(9(15)5-7)17-12-4-2-8(14)6-10(12)16/h1-6,16H

InChI key

XEFQLINVKFYRCS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

A component of cefsulodin-irgasan-novobiocin selection medium for Yersinia, a human pathogen that may contaminate animal-source food products.

Biochem/physiol Actions

It is an inhibitor of the enoyl-ACP (acyl-carrier protein) reductase component of type II fatty acid synthase (FAS-II) in bacteria and Plasmodium. It also inhibits mammalian fatty acid synthase (FASN), and may have anticarcinogenic activity.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Narasimha Pullaguri et al.
Environmental toxicology and pharmacology, 75, 103311-103311 (2019-12-17)
Triclosan is widely used in consumer products as an antimicrobial agent. Epidemiological studies have reported the association of triclosan with adverse birth outcomes. The toxic effects of triclosan on the developing stages of zebrafish are reported, however, its role as
Gokce Kor-Bicakci et al.
Molecules (Basel, Switzerland), 25(2) (2020-01-17)
Treatment of emerging contaminants, such as antimicrobials, has become a priority topic for environmental protection. As a persistent, toxic, and bioaccumulative antimicrobial, the accumulation of triclosan (TCS) in wastewater sludge is creating a potential risk to human and ecosystem health
Sunjin Lee et al.
Ecotoxicology and environmental safety, 182, 109377-109377 (2019-06-30)
The measurement of oxygen consumption rate (OCR) provides a comprehensive understanding of mitochondrial metabolism. However, no study has been conducted to investigate the mitochondrial dysfunction caused by organophosphate flame retardants (OPFRs). The objectives of this study were to optimize the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service