Skip to Content
Merck
All Photos(1)

Key Documents

93105

Supelco

Avenanthramide B

analytical standard

Synonym(s):

5-Hydroxy-2-{[(2E)-3-(4-hydroxy-3-methoxyphenyl)-1-oxo-2-propenyl]amino}benzoic acid, N-[4′-Hydroxy-3′-methoxy-(E)-cinnamoyl]-5-hydroxyanthranilic acid, Avenanthramide Bf

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$819.00

$819.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$819.00

About This Item

Empirical Formula (Hill Notation):
C17H15NO6
Molecular Weight:
329.30
Beilstein:
3625397
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$819.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

SMILES string

OC1=CC=C(NC(/C=C/C2=CC=C(O)C(OC)=C2)=O)C(C(O)=O)=C1

InChI

1S/C17H15NO6/c1-24-15-8-10(2-6-14(15)20)3-7-16(21)18-13-5-4-11(19)9-12(13)17(22)23/h2-9,19-20H,1H3,(H,18,21)(H,22,23)/b7-3+

InChI key

JXFZHMCSCYADIX-XVNBXDOJSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhuohong Xie et al.
Food chemistry, 224, 280-288 (2017-02-06)
Avenanthramides (AVNs) are a family of nitrogen-containing phenolic compounds produced in oat; AVN 2c, 2p, and 2f are the three major members. An LC-MS/MS method was developed, with the limit of detection (LOD) and the limit of quantitation (LOQ) being
Oswaldo Hernandez-Hernandez et al.
Food chemistry, 343, 128408-128408 (2020-11-08)
From a mutagenized oat population, produced by ethyl methanesulfonate mutagenesis, hulled grains from 17 lines with elevated avenanthramide (AVN) content were selected and their AVN structures, concentrations and antioxidant potentials were determined by HPLC-MS2 and HPLC equipped with an on-line

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service