30366
Avenanthramide A
analytical standard
Synonym(s):
5-Hydroxy-2-{[(2E)-3-(4-hydroxyphenyl)-1-oxo-2-propenyl]amino}benzoic acid, N-(4′-Hydroxy-(E)-cinnamoyl)-5-hydroxyanthranilic acid, Avenanthramide Bp
Select a Size
About This Item
Recommended Products
grade
analytical standard
Quality Level
Assay
≥98.0% (HPLC)
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
format
neat
storage temp.
2-8°C
SMILES string
OC1=CC=C(NC(/C=C/C2=CC=C(O)C=C2)=O)C(C(O)=O)=C1
InChI
1S/C16H13NO5/c18-11-4-1-10(2-5-11)3-8-15(20)17-14-7-6-12(19)9-13(14)16(21)22/h1-9,18-19H,(H,17,20)(H,21,22)/b8-3+
InChI key
QGUMNWHANDITDB-FPYGCLRLSA-N
Application
Recommended products
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service