Skip to Content
Merck
All Photos(3)

Key Documents

92427

Supelco

Bisphenol A (3-chloro-2-hydroxypropyl) (2,3-dihydroxypropyl) ether

analytical standard

Synonym(s):

2-[4-(3-Chloro-2-hydroxypropyloxy)phenyl]-2-[4-(2,3-dihydroxypropyloxy)phenyl]propane, BADGE-HCl-H2O

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$310.00
100 MG
$907.00

$310.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$310.00
100 MG
$907.00

About This Item

Empirical Formula (Hill Notation):
C21H27ClO5
CAS Number:
Molecular Weight:
394.89
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$310.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CC(C1=CC=C(OCC(CCl)O)C=C1)(C)C2=CC=C(OCC(O)CO)C=C2

InChI

1S/C21H27ClO5/c1-21(2,15-3-7-19(8-4-15)26-13-17(24)11-22)16-5-9-20(10-6-16)27-14-18(25)12-23/h3-10,17-18,23-25H,11-14H2,1-2H3

InChI key

HDTYUHNZRYZEEB-UHFFFAOYSA-N

General description

Bisphenol A (3-chloro-2-hydroxypropyl) (2,3-dihydroxypropyl) ether is a derivative of Bisphenol A diglycidyl ether and an exogenous endocrine disruptor widely present in canned food samples.[1]

Application

Bisphenol A (3-chloro-2-hydroxypropyl) (2,3-dihydroxypropyl) ether may be used as an analytical reference standard for the quantification of the analyte in food cans using high performance chromatography-tandem mass spectrometry technique.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Standard for determination of toxic BADGE and derivatives in canned food[2][3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of bisphenol A diglycidyl ether and its hydrolysis and chlorohydroxy derivatives by liquid chromatography?mass spectrometry
Garcia SR and Losado PP
Journal of Chromatography A, 1032(1-2), 37-43 (2004)
J E Biles et al.
Journal of agricultural and food chemistry, 47(5), 1965-1969 (1999-12-20)
Migration of the diglycidyl ether of bisphenol A (DGEBA) to food from can enamels and can pull-top seals is reported. Derivatives of DGEBA are also determined in some foods. Levels of DGEBA in the foods surveyed in this study range
M. Biedermann et al.
Mitteilungen Aus Dem Gebiete Der Lebensmittel-Untersuchung Un Hygiene = Travaux De Chimie Alimentaire Et D'Hygiene, 90, 177-177 (1999)
Xinhua Wu et al.
Se pu = Chinese journal of chromatography, 28(11), 1094-1098 (2011-03-09)
A comprehensive analytical method based on high performance liquid chromatography-tandem mass spectrometry (HPLC-MS/MS) was developed for measuring 6 exogenous endocrine disruptors--bisphenol diglycidyl ethers, including bisphenol A diglycidyl ether (BADGE), bisphenol A glycidyl (2,3-dihydroxypropyl) ether (BADGE x H2O), bisphenol A glycidyl
Natalia Szczepańska et al.
The Science of the total environment, 708, 135178-135178 (2019-12-04)
The problem of the presence of trace organic pollutants in food is of growing importance due to increasing awareness about their impact on newborns, infants and adults of reproductive age. Despite the fact that packaged food products offer many advantages

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service