Skip to Content
Merck
All Photos(1)

Key Documents

90477

Supelco

Bisphenol AF

Bisphenol AF

analytical standard

Synonym(s):

4,4′-(Hexafluoroisopropylidene)diphenol, 2,2-Bis(4-hydroxyphenyl)hexafluoropropane, Bisphenol AF, Hexafluorobisphenol A

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$91.60

$91.60


Estimated to ship onMay 19, 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$91.60

About This Item

Linear Formula:
(CF3)2C(C6H4OH)2
CAS Number:
Molecular Weight:
336.23
Beilstein:
1891568
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$91.60


Estimated to ship onMay 19, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

160-163 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

Oc1ccc(cc1)C(c2ccc(O)cc2)(C(F)(F)F)C(F)(F)F

InChI

1S/C15H10F6O2/c16-14(17,18)13(15(19,20)21,9-1-5-11(22)6-2-9)10-3-7-12(23)8-4-10/h1-8,22-23H

InChI key

ZFVMWEVVKGLCIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service