Skip to Content
Merck
All Photos(1)

Key Documents

92337

Sigma-Aldrich

Bromotrimethylsilane

purum, ≥97.0% (AT)

Synonym(s):

TMBS, Trimethylbromosilane, Trimethylsilyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiBr
CAS Number:
Molecular Weight:
153.09
Beilstein:
1731135
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

grade

purum

Quality Level

Assay

≥97.0% (AT)

form

liquid

contains

silver wool as stabilizer

refractive index

n20/D 1.424 (lit.)
n20/D 1.424

bp

79 °C (lit.)

density

1.16 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C[Si](C)(C)Br

InChI

1S/C3H9BrSi/c1-5(2,3)4/h1-3H3

InChI key

IYYIVELXUANFED-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bromotrimethylsilane is used with Cl3 to catalyze the direct allylation of a variety of alcohols with allyltrimethylsilane.

Application

Bromotrimethylsilane may be used as a derivatizing reagent for the determination of alkyl lysophospholipids in cell culture media using gas chromatography technique.[1]

Other Notes

Powerful silylating agent [2][3]; Cleavage of ethers [4]; Smooth cleavage of phosphonic and phosphoric alkyl esters [5]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C. Flann et al.
Journal of the American Chemical Society, 109, 6097-6097 (1987)
J. Heberle et al.
Silylating Agents, 2nd ed. (1995)
J.L. Meek et al.
Journal of the American Chemical Society, 110, 2317-2317 (1988)
S. Hanessian et al.
Tetrahedron Letters, 25, 2515-2515 (1984)
Gas chromatographic determination of alkyl lysophospholipids after solid-phase extraction from cell culture media
Coene J, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 612(1), 21-26 (1993)

Articles

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service