Skip to Content
Merck
All Photos(2)

Key Documents

92167

Supelco

Oleuropein

analytical standard

Synonym(s):

(2S,3E,4S)-3-Ethylidene-2-(β-D-glucopyranosyloxy)-3,4-dihydro-5-(methoxycarbonyl)-2H-pyran-4-acetic acid 2-(3,4-dihydroxyphenyl)ethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$241.00

$241.00


Estimated to ship onApril 11, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$241.00

About This Item

Empirical Formula (Hill Notation):
C25H32O13
CAS Number:
Molecular Weight:
540.51
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$241.00


Estimated to ship onApril 11, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤5.0% water

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

COC(=O)C1=CO[C@@H](O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)\C(=C\C)[C@@H]1CC(=O)OCCc3ccc(O)c(O)c3

InChI

1S/C25H32O13/c1-3-13-14(9-19(29)35-7-6-12-4-5-16(27)17(28)8-12)15(23(33)34-2)11-36-24(13)38-25-22(32)21(31)20(30)18(10-26)37-25/h3-5,8,11,14,18,20-22,24-28,30-32H,6-7,9-10H2,1-2H3/b13-3+/t14-,18+,20+,21-,22+,24-,25-/m0/s1

InChI key

RFWGABANNQMHMZ-ZCHJGGQASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Oleuropein is a phenolic compound with pungent odor, which is found in extra virgin olive oil.[1] It can undergo hydrolysis by Lactobacillus plantarum via action of β-glucosidase and esterase to form elenolic acid and hydroxytyrosol as the final products.[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oleuropein aglycone enhances UCP1 expression in brown adipose tissue in high-fat-diet-induced obese rats by activating ?-adrenergic signaling
Oi-Kano Y, et al.
The Journal of Nutritional Biochemistry, 40, 209-218 (2017)
Oleuropein hydrolysis by lactic acid bacteria in natural green olives
Ramirez E, et al.
LWT--Food Science and Technology, 78, 165-171 (2017)
Ali Reza Khalatbary et al.
Neurological research, 34(1), 44-51 (2011-12-27)
Oleuropein (OE) is a well-known antioxidant polyphenol from olive oil. The purpose of this study was to determine the potential neuroprotective effects of oleuropein in an experimental spinal cord injury model. Rats were randomly divided into four groups of 21
Ali Reza Khalatbary et al.
Iranian biomedical journal, 15(4), 164-167 (2012-03-08)
Neutrophil infiltration plays an important role in inflammatory reactions following spinal cord injury (SCI) and these cells cause substantial secondary tissue damage. The purpose of this study was to determine the effect of oleuropein (OE) on myeloperoxidase (MPO) activity as
S Fernández-Arroyo et al.
Journal of pharmaceutical and biomedical analysis, 63, 128-134 (2012-03-01)
Crude phenolic extracts (PE) have been obtained from naturally bearing Spanish extra-virgin olive oil (EVOO) showing different polyphenol families such as secoiridoids, phenolic alcohols, lignans, and flavones. EVOO-derived complex phenols (especially from the Arbequina variety olive) have been shown to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service