Skip to Content
Merck
All Photos(1)

Key Documents

91214

Supelco

Isofraxidin

analytical standard

Synonym(s):

6,8-Dimethoxyumbelliferone, 7-Hydroxy-6,8-dimethoxychromen-2-one, 7-Hydroxy-6,8-dimethoxycoumarin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10O5
CAS Number:
Molecular Weight:
222.19
Beilstein:
202652
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤5.0% water

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

COc1cc2C=CC(=O)Oc2c(OC)c1O

InChI

1S/C11H10O5/c1-14-7-5-6-3-4-8(12)16-10(6)11(15-2)9(7)13/h3-5,13H,1-2H3

InChI key

HOEVRHHMDJKUMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isofraxidin is a major active constituent, isolated from Sarcandra glabra plant with a variety of biological properties such as antifatigue, antistress, anticancer, immunoaccomondating and cholagogic effects.[1]

Application

Isofraxidin may be used as an analytical reference standard for the separartion and quantification of the analyte in Sarcandra glabra plant[1] and Acanthopanax senticosus Harms[2] using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiaqin Liu et al.
Bioorganic & medicinal chemistry, 12(2), 469-474 (2004-01-16)
This study was designed to examine the interaction of isofraxidin with human serum albumin (HSA) under physiological conditions with drug concentrations in the range of 3.3 x 10(-6) mol L(-1)-3.0x10(-5) mol L(-1) and HSA concentration at 1.5 x 10(-6) mol
Jiaqin Liu et al.
Biopolymers, 73(4), 443-450 (2004-03-03)
The binding of isofraxidin to bovine serum albumin (BSA) was studied under physiological conditions with BSA concentration of 1.5 x 10(-6) mol x L(-1) and drug concentration in the range of 1.67 x 10(-6) mol x L(-1) to 2.0 x
Xiaofeng Niu et al.
International immunopharmacology, 14(2), 164-171 (2012-07-18)
Isofraxidin (IF) is a Coumarin compound that can be isolated from medicinal plants, such as Sarcandra glabra (Thunb.). Nakai is widely used in Asian countries for the treatment of anti-bacterial, anti-inflammatory and anti-tumour action. The present investigation was designed to
J H Kwak et al.
Journal of natural products, 64(8), 1081-1083 (2001-08-25)
Four new coumarin monoterpene ethers, artekeiskeanols A-D (1-4), and three known coumarins, isofraxidin, fraxidin, and daphnoretin, were isolated from the whole plants of Artemisia keiskeana. All structures were determined from spectral data, and that of artekeiskeanol A (1) was confirmed
T Yamazaki et al.
Toxicology in vitro : an international journal published in association with BIBRA, 21(8), 1530-1537 (2007-06-15)
(+)-Syringaresinol-di-O-beta-D-glucoside (SR), syringin, and isofraxidin isolated from the stem bark of Acanthopanax senticosus Harms are its major constituents. The present work was undertaken to analyze effects of these compounds on inflammatory functions in SW982 human synovial sarcoma cell system. When

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service