Skip to Content
Merck
All Photos(1)

Key Documents

935557

Sigma-Aldrich

3-O-p-Coumaroylquinic acid

≥95%

Synonym(s):

3-O-p-Coumaroylquinic acid, (1S,3R,4R,5R)-1,3,4-Trihydroxy-5-[[3-(4-hydroxyphenyl)-1-oxo-2-propen-1-yl]oxy]cyclohexanecarboxylic acid,, 3-O-p-Coumarylquinic acid, Cinnamic acid, p-hydroxy-, 3-ester with 1,3,4,5-tetrahydroxycyclohexanecarboxylic acid, Cyclohexanecarboxylic acid, 1,3,4-trihydroxy-5-[[3-(4-hydroxyphenyl)-1-oxo-2-propen-1-yl]oxy]-, (1S,3R,4R,5R)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18O8
CAS Number:
Molecular Weight:
338.31
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

Assay

≥95%

form

solid

color

white to light yellow

bp

613.2 °C

density

1.554 g/cm3

storage temp.

2-8°C

SMILES string

O=C([C@]1(O)C[C@@H](O)[C@@H](O)[C@H](OC(/C=C/C2=CC=C(O)C=C2)=O)C1)O

InChI

InChI=1S/C16H18O8/c17-10-4-1-9(2-5-10)3-6-13(19)24-12-8-16(23,15(21)22)7-11(18)14(12)20/h1-6,11-12,14,17-18,20,23H,7-8H2,(H,21,22)/t11-,12-,14-,16+/m1/s1

InChI key

BMRSEYFENKXDIS-OTCYKTEZSA-N

Application

A naturally occurring substance discovered in plants, which possesses antibacterial characteristics and may serve as a substitute for antibiotics in combatting opportunistic fungal infections.

Features and Benefits

The hypoglycemic effects of 3-p-Coumaroylquinic acid are attributed to its capacity to hinder glucose uptake in the intestines.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chlorogenic acid and coffee prevent hypoxia-induced retinal degeneration
Jang Holim, et. al.
Journal of Agricultural and Food Chemistry, 62 (2014)
Roles of chlorogenic Acid on regulating glucose and lipids metabolism: a review
Meng Shengxi, et. al.
Evidence-Based Complementary and Alternative Medicine : ECAM (2013)
Chlorogenic acid and caffeic acid are absorbed in humans
M R Olthof, et. al.
The Journal of Nutrition, 131 (2001)
Computational screen and experimental validation of anti-influenza effects of quercetin and chlorogenic acid from traditional Chinese medicine
Liu Zekun, et. al.
Scientific reports, 6 (2016)
Holim Jang et al.
Journal of agricultural and food chemistry, 62(1), 182-191 (2013-12-04)
This study explored whether chlorogenic acid (CGA) and coffee have protective effects against retinal degeneration. Under hypoxic conditions, the viability of transformed retinal ganglion (RGC-5) cells was significantly reduced by treatment with the nitric oxide (NO) donor S-nitroso-N-acetylpenicillamine (SNAP). However

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service